Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000430888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000430888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000444900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
ENST00000521074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010726.hg.1 |
NR_024340.1 |
Refseq |