Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05001973_m1
          See other GSTO2 GE Assays ›
          Gene Symbol:
          GSTO2
          Gene Name:
          glutathione S-transferase omega 2
          Gene Aliases:
          GSTO 2-2, bA127L20.1
          Chromosome Location:
          Chr.10: 104267626 - 104304948 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05001973_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GSTO2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191014.1 NP_001177943.1 2 - 3 429 123
          NM_183239.1 NP_899062.1 4 - 5 994 123
          XM_006717624.3 XP_006717687.1 4 - 5 683 123
          XM_011539270.2 XP_011537572.1 4 - 5 2057 123
          XM_011539271.1 XP_011537573.1 4 - 5 683 123
          XM_011539272.2 XP_011537574.1 4 - 5 683 123
          GenBank mRNA AK291886.1 - 4 - 5 682 123
          AK300508.1 - 3 - 4 391 123
          AY191318.1 - 3 - 4 362 123
          AY209189.1 - 4 - 5 686 123
          AY350731.1 - 4 - 5 686 123
          BC023583.2 - 2 - 3 395 123
          BC056918.1 - 4 - 5 994 5165084 123

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GSTO2

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          xenobiotic metabolic process
          L-ascorbic acid metabolic process
          cellular response to arsenic-containing substance
          cellular oxidant detoxification
          glutathione derivative biosynthetic process
          glutathione transferase activity
          protein binding
          oxidoreductase activity
          glutathione dehydrogenase (ascorbate) activity
          methylarsonate reductase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000498052.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000498052.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000498052.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 ENST00000498052.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000338595.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000450629.6 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000467629.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000473401.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000477078.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000498052.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000498052.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000498052.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 ENST00000498052.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191013.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191014.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_001191015.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012498.hg.1 NM_183239.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0