Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000611481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000616974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
ENST00000631321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006555.hg.1 |
NR_103810.1 |
Refseq |