Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05008365_s1
          See other SIPA1L1 GE Assays ›
          Gene Symbol:
          SIPA1L1
          Gene Name:
          signal induced proliferation associated 1 like 1
          Gene Aliases:
          E6TP1
          Chromosome Location:
          Chr.14: 71320418 - 71741229 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05008365_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SIPA1L1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001284245.1 NP_001271174.1 21 7107 102
          NM_001284246.1 NP_001271175.1 21 7693 102
          NM_001284247.1 NP_001271176.1 21 7108 102
          NM_015556.2 NP_056371.1 22 7170 102
          XM_005267516.4 XP_005267573.1 22 7759 102
          XM_006720111.2 XP_006720174.1 24 9046 102
          XM_011536630.1 XP_011534932.1 26 7521 102
          XM_011536631.1 XP_011534933.1 26 7932 102
          XM_011536632.1 XP_011534934.1 25 7441 102
          XM_011536635.2 XP_011534937.1 25 7588 102
          XM_011536638.2 XP_011534940.1 21 6244 102
          XM_017021159.1 XP_016876648.1 28 7678 102
          XM_017021160.1 XP_016876649.1 26 7600 102
          XM_017021161.1 XP_016876650.1 27 7553 102
          XM_017021162.1 XP_016876651.1 29 7735 102
          XM_017021163.1 XP_016876652.1 24 7402 102
          XM_017021164.1 XP_016876653.1 24 7409 102
          XM_017021165.1 XP_016876654.1 26 7478 102
          XM_017021166.1 XP_016876655.1 27 7611 102
          XM_017021167.1 XP_016876656.1 26 7486 102
          XM_017021168.1 XP_016876657.1 26 7460 102
          XM_017021169.1 XP_016876658.1 25 7613 102
          XM_017021170.1 XP_016876659.1 25 7971 102
          XM_017021171.1 XP_016876660.1 26 7544 102
          XM_017021172.1 XP_016876661.1 25 7419 102
          XM_017021173.1 XP_016876662.1 24 7546 102
          XM_017021174.1 XP_016876663.1 24 7786 102
          XM_017021175.1 XP_016876664.1 24 7904 102
          XM_017021176.1 XP_016876665.1 29 7731 102
          XM_017021177.1 XP_016876666.1 29 7720 102
          XM_017021178.1 XP_016876667.1 27 7558 102
          XM_017021179.1 XP_016876668.1 26 7461 102
          XM_017021180.1 XP_016876669.1 30 7773 102
          XM_017021181.1 XP_016876670.1 28 7675 102
          XM_017021182.1 XP_016876671.1 27 7608 102
          XM_017021183.1 XP_016876672.1 25 7416 102
          XM_017021184.1 XP_016876673.1 29 7728 102
          XM_017021185.1 XP_016876674.1 22 7756 102
          XM_017021186.1 XP_016876675.1 27 7615 102
          XM_017021187.1 XP_016876676.1 25 7481 102
          XM_017021188.1 XP_016876677.1 24 7356 102
          XM_017021189.1 XP_016876678.1 26 7465 102
          XM_017021190.1 XP_016876679.1 27 7612 102
          XM_017021191.1 XP_016876680.1 25 7420 102
          XM_017021192.1 XP_016876681.1 24 7353 102
          XM_017021193.1 XP_016876682.1 24 7351 102
          XM_017021194.1 XP_016876683.1 27 7531 102
          XM_017021195.1 XP_016876684.1 26 7468 102
          XM_017021196.1 XP_016876685.1 26 7465 102
          XM_017021197.1 XP_016876686.1 27 5878 102
          XM_017021198.1 XP_016876687.1 24 5619 102
          XM_017021199.1 XP_016876688.1 28 5920 102
          XM_017021200.1 XP_016876689.1 24 5620 102
          GenBank mRNA BC033950.2 - 1 218 102

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SIPA1L1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          actin cytoskeleton reorganization
          regulation of GTPase activity
          ephrin receptor signaling pathway
          regulation of synaptic plasticity
          regulation of axonogenesis
          regulation of small GTPase mediated signal transduction
          regulation of dendritic spine morphogenesis
          activation of GTPase activity
          molecular_function
          GTPase activator activity
          ephrin receptor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc