Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05010174_g1
          See other RSL24D1 GE Assays ›
          Gene Symbol:
          RSL24D1
          Gene Name:
          ribosomal L24 domain containing 1
          Gene Aliases:
          C15orf15, HRP-L30-iso, L30, RLP24, RPL24, RPL24L, TVAS3
          Chromosome Location:
          Chr.15: 55181314 - 55197033 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05010174_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RSL24D1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016304.2 NP_057388.1 4 - 5 473 111
          GenBank mRNA AB462502.1 - 4 - 5 357 111
          AF060926.1 - 4 - 5 340 111
          AF165521.1 - 4 - 5 473 111
          AF201949.1 - 4 - 5 473 111
          AF212226.1 - 4 - 5 473 111
          AK055713.1 - 4 - 5 360 111
          AK315431.1 - 4 - 5 373 111
          BC008409.1 - 4 - 5 395 4097130 111
          BC008422.1 - 4 - 5 365 4096889 111
          BC008449.1 - 4 - 5 371 4133589 111
          BC008499.1 - 4 - 5 386 4286115 111
          BC009593.1 - 4 - 5 376 4109097 111
          BC009604.1 - 4 - 5 376 4282059 111
          BC012913.1 - 4 - 5 340 4514979 111
          BC014576.1 - 4 - 5 375 3930303 111
          BC016312.1 - 4 - 5 381 4096867 111
          BC016331.1 - 4 - 5 381 4108672 111
          BC016725.1 - 4 - 5 382 4078305 111
          BC016777.1 - 4 - 5 363 4104810 111
          BC026266.1 - 4 - 5 377 4276092 111
          BC026267.1 - 4 - 5 356 4277062 111
          BC028672.1 - 4 - 5 362 4092804 111
          BC035995.1 - 4 - 5 386 4671856 111
          BC093067.1 - 4 - 5 360 30419021 111
          CR457179.1 - 4 - 5 330 111

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RSL24D1

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal large subunit assembly
          translation
          assembly of large subunit precursor of preribosome
          structural constituent of ribosome
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200012781.hg.1 ENST00000445584.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011849.hg.1 ENST00000509365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011330.hg.1 ENST00000449255.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800011302.hg.1 ENST00000524051.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562895.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000564344.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000564344.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000564344.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000564344.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007405.hg.1 ENST00000591045.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200008611.hg.1 ENST00000604494.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007762.hg.1 ENST00000564321.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007484.hg.1 ENST00000556832.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008575.hg.1 ENST00000549019.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600011221.hg.1 ENST00000404972.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800009389.hg.1 ENST00000518519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800008711.hg.1 ENST00000578420.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007064.hg.1 ENST00000434442.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012191.hg.1 ENST00000400938.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013548.hg.1 ENST00000615161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013548.hg.1 ENST00000615161.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008842.hg.1 ENST00000426934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011059.hg.1 ENST00000553036.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010446.hg.1 ENST00000605454.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010446.hg.1 ENST00000605454.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000260443.8 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562895.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562895.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562895.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562895.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000562993.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000564344.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000564344.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000564344.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000564344.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000565854.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 ENST00000569386.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC1500009492.hg.1 NM_016304.2 Refseq
          Affymetrix Human Clariom D Assay TC0100012191.hg.1 ENST00000400938.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600011221.hg.1 ENST00000404972.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200008575.hg.1 ENST00000549019.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011330.hg.1 ENST00000449255.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007762.hg.1 ENST00000564321.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013548.hg.1 ENST00000615161.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013548.hg.1 ENST00000615161.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011059.hg.1 ENST00000553036.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007484.hg.1 ENST00000556832.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800008711.hg.1 ENST00000578420.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800011302.hg.1 ENST00000524051.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008842.hg.1 ENST00000426934.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012781.hg.1 ENST00000445584.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011849.hg.1 ENST00000509365.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800009389.hg.1 ENST00000518519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200008611.hg.1 ENST00000604494.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007064.hg.1 ENST00000434442.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007405.hg.1 ENST00000591045.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0