Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05017467_s1
          See other ICAM1 GE Assays ›
          Gene Symbol:
          ICAM1
          Gene Name:
          intercellular adhesion molecule 1
          Gene Aliases:
          BB2, CD54, P3.58
          Chromosome Location:
          Chr.19: 10270841 - 10286615 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05017467_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ICAM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000201.2 NP_000192.2 7 2235 95
          GenBank mRNA AK130659.1 - 7 1956 95
          BC015969.2 - 7 1987 3506766 95
          J03132.1 - 7 1973 95
          M24283.1 - 7 1987 95
          M55091.1 - 1 121 95

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ICAM1

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          regulation of leukocyte mediated cytotoxicity
          response to amphetamine
          T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          acute inflammatory response to antigenic stimulus
          T cell antigen processing and presentation
          positive regulation of cellular extravasation
          cell adhesion
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          leukocyte cell-cell adhesion
          cell aging
          sensory perception of sound
          regulation of cell shape
          response to xenobiotic stimulus
          response to ionizing radiation
          response to sulfur dioxide
          response to organic cyclic compound
          membrane to membrane docking
          regulation of cell adhesion
          extracellular matrix organization
          positive regulation of actin filament polymerization
          cellular response to nutrient levels
          cell adhesion mediated by integrin
          response to gonadotropin
          response to amino acid
          positive regulation of GTPase activity
          adhesion of symbiont to host
          positive regulation of nitric oxide biosynthetic process
          response to ethanol
          positive regulation of vasoconstriction
          response to copper ion
          viral entry into host cell
          receptor-mediated virion attachment to host cell
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of immune response
          leukocyte migration
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of calcium ion transport
          interferon-gamma-mediated signaling pathway
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to alkaloid
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to hypoxia
          establishment of Sertoli cell barrier
          regulation of ruffle assembly
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell apoptotic process
          virus receptor activity
          transmembrane signaling receptor activity
          integrin binding
          protein binding
          signaling receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000264832.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000423829.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000585443.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000588645.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 ENST00000592686.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq
          Affymetrix Human Clariom D Assay TC1900006977.hg.1 NM_000201.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc