Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05020047_g1
          See other APOA2 GE Assays ›
          Gene Symbol:
          APOA2
          Gene Name:
          apolipoprotein A2
          Gene Aliases:
          Apo-AII, ApoA-II, apoAII
          Chromosome Location:
          Chr.1: 161222293 - 161223628 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05020047_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: APOA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001643.1 NP_001634.1 3 - 4 245 88
          GenBank mRNA AK312034.1 - 3 - 4 241 88
          BC005282.1 - 3 - 4 245 3934476 88
          BT006786.1 - 2 - 3 187 88
          M29882.1 - 2 - 3 196 88
          X00955.1 - 2 - 3 238 88

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene APOA2

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          acute inflammatory response
          triglyceride metabolic process
          phosphatidylcholine biosynthetic process
          cholesterol metabolic process
          phospholipid catabolic process
          response to xenobiotic stimulus
          response to glucose
          positive regulation of cholesterol esterification
          negative regulation of very-low-density lipoprotein particle remodeling
          viral process
          protein oxidation
          peptidyl-methionine modification
          regulation of intestinal cholesterol absorption
          animal organ regeneration
          regulation of protein stability
          negative regulation of cholesterol transport
          cholesterol efflux
          phospholipid efflux
          triglyceride-rich lipoprotein particle remodeling
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          high-density lipoprotein particle clearance
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          cholesterol homeostasis
          positive regulation of catalytic activity
          response to estrogen
          reverse cholesterol transport
          diacylglycerol catabolic process
          negative regulation of lipid catabolic process
          positive regulation of lipid catabolic process
          response to glucocorticoid
          negative regulation of lipase activity
          negative regulation of cholesterol import
          negative regulation of cholesterol transporter activity
          lipid transporter activity
          protein binding
          phospholipid binding
          high-density lipoprotein particle binding
          lipid binding
          cholesterol binding
          phosphatidylcholine binding
          apolipoprotein receptor binding
          protein homodimerization activity
          protein heterodimerization activity
          lipase inhibitor activity
          phosphatidylcholine-sterol O-acyltransferase activator activity
          high-density lipoprotein particle receptor binding
          cholesterol transfer activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000469730.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000469730.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000469730.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000469730.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000491350.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000491350.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000491350.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 ENST00000491350.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000367990.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463273.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000463812.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000464492.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000468465.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000469730.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000469730.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000469730.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000469730.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000470459.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481413.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000481511.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000491350.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000491350.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000491350.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 ENST00000491350.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 NM_001643.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016166.hg.1 NM_001643.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0