Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05020091_m1
          See other CHRM3 GE Assays ›
          Gene Symbol:
          CHRM3
          Gene Name:
          cholinergic receptor muscarinic 3
          Gene Aliases:
          EGBRS, HM3, PBS
          Chromosome Location:
          Chr.1: 239386565 - 239915450 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05020091_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CHRM3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000740.2 NP_000731.1 4 - 5 764 157
          XM_005273032.3 XP_005273089.1 5 - 6 1069 157
          XM_011544041.2 XP_011542343.1 6 - 7 1168 157
          XM_011544043.2 XP_011542345.1 8 - 9 1501 157
          XM_011544044.2 XP_011542346.1 7 - 8 1272 157
          XM_011544045.2 XP_011542347.1 7 - 8 1397 157
          XM_011544046.2 XP_011542348.1 5 - 6 1105 157
          XM_017000152.1 XP_016855641.1 7 - 8 4094 157
          XM_017000153.1 XP_016855642.1 6 - 7 1172 157
          XM_017000154.1 XP_016855643.1 5 - 6 866 157
          XM_017000155.1 XP_016855644.1 6 - 7 1209 157
          XM_017000156.1 XP_016855645.1 6 - 7 1173 157
          XM_017000157.1 XP_016855646.1 4 - 5 774 157
          XM_017000158.1 XP_016855647.1 4 - 5 1006 157
          XM_017000160.1 XP_016855649.1 3 - 4 1828 157
          XM_017000161.1 XP_016855650.1 5 - 6 1110 157
          XM_017000163.1 XP_016855652.1 3 - 4 4804 157
          GenBank mRNA AF279779.1 - 4 - 5 763 157
          BC121026.2 - 1 - 2 128 40117429 157
          BC121027.2 - 1 - 2 157

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF279779.1, BC121026.2, NM_000740.2, XM_005273032.3, XM_011544041.2, XM_011544043.2, XM_011544044.2, XM_011544045.2, XM_011544046.2, XM_017000152.1, XM_017000153.1, XM_017000154.1, XM_017000155.1, XM_017000156.1, XM_017000157.1, XM_017000158.1, XM_017000160.1, XM_017000161.1, XM_017000163.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CHRM3

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of vascular associated smooth muscle contraction
          cellular protein modification process
          smooth muscle contraction
          signal transduction
          G protein-coupled receptor signaling pathway
          adenylate cyclase-inhibiting G protein-coupled acetylcholine receptor signaling pathway
          phospholipase C-activating G protein-coupled acetylcholine receptor signaling pathway
          G protein-coupled acetylcholine receptor signaling pathway
          synaptic transmission, cholinergic
          nervous system development
          cell population proliferation
          positive regulation of smooth muscle contraction
          saliva secretion
          phosphatidylinositol phospholipase C activity
          protein binding
          G protein-coupled acetylcholine receptor activity
          signaling receptor activity
          acetylcholine binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000448020.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000448020.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000448020.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000487470.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000487470.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000487470.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000255380.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000448020.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000448020.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000448020.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000487470.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000487470.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000487470.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 ENST00000615928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018367.hg.1 NM_000740.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0