Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05020518_g1
          See other POU5F1P4 GE Assays ›
          Gene Symbol:
          POU5F1P4
          Gene Name:
          POU class 5 homeobox 1 pseudogene 4
          Gene Aliases:
          Oct4-pg4, POU5FLC1
          Chromosome Location:
          Chr.1: 155433180 - 155434262 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05020518_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: POU5F1P4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034180.1 1 385 79
          GenBank mRNA AF268613.1 - 1 385 79
          DQ851566.1 - 1 385 79
          FM178001.1 - 1 263 79
          FM178003.1 - 1 263 79
          FM178004.1 - 1 263 79
          FM178009.1 - 1 263 79
          FM178010.1 - 1 263 79
          GU480814.1 - 1 386 79
          GU480831.1 - 1 387 79
          GU480834.1 - 1 386 79
          GU480836.1 - 1 386 79
          GU480839.1 - 1 386 79
          GU480848.1 - 1 386 79
          GU480853.1 - 1 387 79
          GU480854.1 - 1 386 79
          GU480855.1 - 1 384 79
          GU480857.1 - 1 386 79
          GU480858.1 - 1 386 79
          GU480859.1 - 1 386 79
          GU480860.1 - 1 385 79
          GU480861.1 - 1 387 79
          GU480862.1 - 1 386 79
          GU480863.1 - 1 386 79
          GU480865.1 - 1 387 79
          GU480866.1 - 1 386 79
          GU480867.1 - 1 386 79
          GU480869.1 - 1 386 79
          GU480870.1 - 1 386 79
          GU480871.1 - 1 386 79
          GU480872.1 - 1 386 79
          GU480873.1 - 1 386 79
          GU480874.1 - 1 386 79
          GU480875.1 - 1 386 79
          GU480879.1 - 1 386 79
          GU480881.1 - 1 386 79
          GU480882.1 - 1 386 79
          GU480890.1 - 1 385 79
          GU480895.1 - 1 386 79
          GU480898.1 - 1 385 79
          GU480900.1 - 1 385 79
          GU480904.1 - 1 386 79
          GU480914.1 - 1 385 79
          HQ122676.1 - 1 386 79
          HQ122677.1 - 1 386 79
          HQ122678.1 - 1 384 79
          HQ122679.1 - 1 386 78
          HQ122680.1 - 1 386 79
          HQ122681.1 - 1 386 79
          HQ122682.1 - 1 386 79
          HQ122683.1 - 1 387 79
          HQ122685.1 - 1 384 79
          HQ122686.1 - 1 386 79
          HQ122687.1 - 1 386 79
          HQ122688.1 - 1 385 79
          HQ122689.1 - 1 384 79
          HQ122690.1 - 1 386 79
          HQ122695.1 - 1 385 79
          HQ122696.1 - 1 385 79
          HQ122698.1 - 1 386 79
          HQ122699.1 - 1 386 79
          HQ122700.1 - 1 386 79
          HQ122702.1 - 1 386 79
          HQ122703.1 - 1 386 79
          HQ122705.1 - 1 386 79
          HQ122706.1 - 1 386 79
          HQ122707.1 - 1 385 79
          HQ122708.1 - 1 386 79
          HQ122731.1 - 1 386 79
          HQ122733.1 - 1 386 79
          HQ122735.1 - 1 386 79
          HQ122740.1 - 1 386 79
          HQ122743.1 - 1 386 79
          HQ122745.1 - 1 386 79
          HQ122746.1 - 1 386 79
          HQ122747.1 - 1 386 79
          HQ122748.1 - 1 386 79
          HQ122757.1 - 1 386 79
          HQ122758.1 - 1 386 79
          HQ122760.1 - 1 386 79
          HQ122761.1 - 1 386 79
          HQ122762.1 - 1 386 79
          HQ122763.1 - 1 386 79
          HQ122764.1 - 1 386 79
          HQ122765.1 - 1 386 79
          HQ122766.1 - 1 386 79
          HQ122767.1 - 1 386 79
          HQ122768.1 - 1 386 79
          HQ122769.1 - 1 385 79
          HQ122771.1 - 1 386 79
          HQ122772.1 - 1 386 79
          HQ122774.1 - 1 386 79
          HQ122775.1 - 1 386 79
          HQ122777.1 - 1 385 79
          HQ122778.1 - 1 386 79
          HQ122779.1 - 1 386 79
          HQ122780.1 - 1 386 79
          HQ122798.1 - 1 386 79
          HQ122799.1 - 1 385 79
          HQ122800.1 - 1 387 79
          HQ122801.1 - 1 386 79
          HQ122802.1 - 1 384 79
          HQ122803.1 - 1 387 79
          HQ122804.1 - 1 386 79
          HQ122805.1 - 1 386 79
          HQ122807.1 - 1 386 79
          HQ122808.1 - 1 386 79
          HQ122809.1 - 1 384 79
          HQ122810.1 - 1 386 79
          HQ122811.1 - 1 386 79
          HQ122812.1 - 1 386 79
          HQ122813.1 - 1 386 79
          HQ122814.1 - 1 384 79
          HQ122832.1 - 1 386 79
          HQ122833.1 - 1 386 79
          HQ122834.1 - 1 386 79
          HQ122835.1 - 1 384 79
          HQ122836.1 - 1 386 79
          HQ122837.1 - 1 386 79
          HQ122838.1 - 1 384 79
          HQ122839.1 - 1 386 79
          HQ122840.1 - 1 386 79
          HQ122858.1 - 1 386 79
          HQ122861.1 - 1 386 79
          HQ122862.1 - 1 386 79
          HQ122888.1 - 1 386 79
          HQ122889.1 - 1 386 79
          HQ122890.1 - 1 386 79
          HQ122904.1 - 1 386 79
          HQ122906.1 - 1 386 80
          HQ122907.1 - 1 79
          HQ122908.1 - 1 386 79
          HQ122909.1 - 1 79
          HQ122910.1 - 1 79
          HQ122913.1 - 1 79
          HQ122914.1 - 1 386 79

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100010126.hg.1 ENST00000413175.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010126.hg.1 NR_034180.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010126.hg.1 ENST00000413175.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010126.hg.1 NR_034180.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0