Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
ENST00000451435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
ENST00000451435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
ENST00000451435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
ENST00000451435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
ENST00000479646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
ENST00000479646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
NR_104170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
NR_104170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009917.hg.1 |
NR_104170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
ENST00000451435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
ENST00000451435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
ENST00000451435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
ENST00000451435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
ENST00000479646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
ENST00000479646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
NR_104170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
NR_104170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009917.hg.1 |
NR_104170.1 |
Refseq |