Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05024795_g1
          See other ITSN1 GE Assays ›
          Gene Symbol:
          ITSN1
          Gene Name:
          intersectin 1
          Gene Aliases:
          ITSN, SH3D1A, SH3P17
          Chromosome Location:
          Chr.21: 33642400 - 33899861 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05024795_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITSN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001132.1 NP_001001132.1 28 - 29 3741 86
          NM_003024.2 NP_003015.2 28 - 29 3741 86
          XM_005261026.2 XP_005261083.1 27 - 28 3804 86
          XM_005261027.1 XP_005261084.1 27 - 28 3804 86
          XM_005261031.1 XP_005261088.1 25 - 26 3591 86
          XM_011529691.1 XP_011527993.1 27 - 28 3693 86
          XM_011529692.1 XP_011527994.1 25 - 26 3480 86
          XM_017028428.1 XP_016883917.1 29 - 30 3739 86
          XM_017028429.1 XP_016883918.1 28 - 29 3708 86
          XM_017028430.1 XP_016883919.1 27 - 28 3693 86
          XM_017028431.1 XP_016883920.1 28 - 29 3819 86
          XM_017028432.1 XP_016883921.1 27 - 28 3804 86
          XM_017028433.1 XP_016883922.1 26 - 27 3606 86
          XM_017028434.1 XP_016883923.1 25 - 26 3591 86
          XM_017028435.1 XP_016883924.1 26 - 27 3495 86
          XM_017028436.1 XP_016883925.1 25 - 26 3480 86
          XM_017028437.1 XP_016883926.1 23 - 24 3159 86
          XM_017028438.1 XP_016883927.1 28 - 29 3708 86
          XM_017028439.1 XP_016883928.1 26 - 27 3606 86
          XM_017028440.1 XP_016883929.1 26 - 27 3495 86
          GenBank mRNA AF064243.1 - 28 - 29 3579 86
          AF064244.1 - 28 - 29 3579 86
          AF114487.1 - 28 - 29 3741 86
          AF114488.1 - 28 - 29 3741 86
          AF180522.1 - 4 - 5 376 86
          AK300274.1 - 27 - 28 3701 86
          AY639648.1 - 3 - 4 289 86
          AY639649.1 - 3 - 4 289 86
          AY639651.1 - 2 - 3 862 86
          AY639652.1 - 3 - 4 407 86
          AY639653.1 - 2 - 3 862 86
          BC116185.1 - 27 - 28 3537 40073780 86
          BC116186.2 - 28 - 29 3552 40073781 86
          BC117560.1 - 25 - 26 3493 7469144 86
          BX538175.1 - 20 - 21 2812 86
          DQ340865.1 - 27 - 28 3537 86
          DQ679754.1 - 25 - 26 3330 86
          DQ679755.1 - 26 - 27 3345 86
          DQ679756.1 - 28 - 29 3432 86
          DQ679757.1 - 27 - 28 3543 86
          EU107386.1 - 28 - 29 3447 86
          EU117382.1 - 25 - 26 3292 86
          EU120733.1 - 26 - 27 3459 86
          EU120734.1 - 26 - 27 3459 86
          EU120735.1 - 25 - 26 3219 86
          EU120736.1 - 24 - 25 3013 86
          EU140799.1 - 26 - 27 3376 86
          EU140800.1 - 25 - 26 3150 86
          EU152331.1 - 23 - 24 3040 86
          EU191901.1 - 26 - 27 3382 86
          GU939616.1 - 20 - 21 2935 86
          GU939617.1 - 19 - 20 2835 86
          U61166.1 - 9 - 10 1409 86

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITSN1

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          regulation of Rho protein signal transduction
          positive regulation of phosphorylation
          positive regulation of apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          ephrin receptor signaling pathway
          synaptic vesicle endocytosis
          regulation of small GTPase mediated signal transduction
          positive regulation of protein kinase B signaling
          guanyl-nucleotide exchange factor activity
          calcium ion binding
          protein binding
          kinase activator activity
          molecular adaptor activity
          proline-rich region binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0