Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05029704_s1
          See other TNIP1 GE Assays ›
          Gene Symbol:
          TNIP1
          Gene Name:
          TNFAIP3 interacting protein 1
          Gene Aliases:
          ABIN-1, NAF1, VAN, nip40-1
          Chromosome Location:
          Chr.5: 151029943 - 151087660 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05029704_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNIP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252385.1 NP_001239314.1 3 426 76
          NM_001252386.1 NP_001239315.1 2 254 76
          NM_001252390.1 NP_001239319.1 3 291 76
          NM_001252391.1 NP_001239320.1 3 323 76
          NM_001252392.1 NP_001239321.1 3 323 76
          NM_001252393.1 NP_001239322.1 3 426 76
          NM_001258454.1 NP_001245383.1 3 426 76
          NM_001258455.1 NP_001245384.1 2 221 76
          NM_001258456.1 NP_001245385.1 2 221 76
          NM_006058.4 NP_006049.3 3 426 76
          XM_005268355.1 XP_005268412.1 3 339 76
          XM_006714751.1 XP_006714814.1 3 339 76
          XM_006714752.2 XP_006714815.1 3 339 76
          XM_017008945.1 XP_016864434.1 3 339 76
          XM_017008946.1 XP_016864435.1 3 305 76
          XM_017008947.1 XP_016864436.1 4 808 76
          XM_017008948.1 XP_016864437.1 2 600 76
          XM_017008949.1 XP_016864438.1 3 339 76
          XM_017008950.1 XP_016864439.1 3 339 76
          XM_017008951.1 XP_016864440.1 2 600 76
          GenBank mRNA AB177543.1 - 2 185 76
          AB177544.1 - 2 185 76
          AB252970.1 - 3 295 76
          AB252971.1 - 3 295 76
          AB252972.1 - 3 416 76
          AB252973.1 - 3 416 76
          AB252974.1 - 2 123 76
          AB252975.1 - 2 244 76
          AB252976.1 - 1 49 76
          AB252977.1 - 1 49 76
          AB252978.1 - 1 49 76
          AB252979.1 - 1 49 76
          AB252980.1 - 1 49 76
          AB252981.1 - 1 49 76
          AB252982.1 - 3 295 76
          AJ011895.1 - 3 295 76
          AJ011896.1 - 3 295 76
          AK074400.1 - 3 357 76
          AK290999.1 - 3 292 76
          AK298685.1 - 2 248 76
          AK299975.1 - 2 186 76
          AK303573.1 - 3 376 76
          AY012155.1 - 3 442 76
          BC012133.1 - 3 332 4549542 76
          BC014008.1 - 3 273 4040118 76
          BP254790.1 - 3 416 76
          BX640647.1 - 2 175 76
          D30755.2 - 2 185 76
          DA643531.1 - 2 151 76

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AB252974.1, AB252975.1, AK299975.1, BX640647.1, NM_001252386.1, XM_017008948.1, XM_017008951.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNIP1

          Gene Ontology Categories:

          Function(s) Process(es)

          MyD88-dependent toll-like receptor signaling pathway
          translation
          proteolysis
          defense response
          inflammatory response
          leukocyte cell-cell adhesion
          glycoprotein biosynthetic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of viral genome replication
          positive regulation of transcription by RNA polymerase II
          positive regulation of inflammatory response
          negative regulation of ERK1 and ERK2 cascade
          modulation by symbiont of host I-kappaB kinase/NF-kappaB cascade
          thiol-dependent deubiquitinase
          protein binding
          mitogen-activated protein kinase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0