Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05030918_gH
          See other CDKN1C GE Assays ›
          Gene Symbol:
          CDKN1C
          Gene Name:
          cyclin dependent kinase inhibitor 1C
          Gene Aliases:
          BWCR, BWS, KIP2, WBS, p57, p57Kip2
          Chromosome Location:
          Chr.11: 2883218 - 2885804 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05030918_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDKN1C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000076.2 NP_000067.1 1 590 93
          NM_001122630.1 NP_001116102.1 2 423 93
          NM_001122631.1 NP_001116103.1 2 423 93
          XM_017017088.1 XP_016872577.1 1 452 93
          GenBank mRNA BC039188.1 - 2 423 93
          BC067842.1 - 1 599 30332904 93
          EU570054.1 - 2 458 93
          U22398.1 - 1 574 93

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDKN1C

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          skeletal system development
          kidney development
          regulation of mitotic cell cycle
          aging
          myeloid cell differentiation
          adrenal gland development
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of kinase activity
          multicellular organism growth
          negative regulation of phosphorylation
          neuron maturation
          camera-type eye development
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of epithelial cell proliferation
          digestive system development
          uterus development
          embryonic placenta morphogenesis
          genetic imprinting
          negative regulation of protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000471157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000471157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000471157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 ENST00000471157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000380725.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000414822.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000430149.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000440480.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000471157.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000471157.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000471157.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 ENST00000471157.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_000076.2 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122630.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122631.1 Refseq
          Affymetrix Human Clariom D Assay TC1100009813.hg.1 NM_001122631.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline