Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05032352_g1
          See other RPL10A GE Assays ›
          Gene Symbol:
          RPL10A
          Gene Name:
          ribosomal protein L10a
          Gene Aliases:
          CSA19, Csa-19, L10A, NEDD6
          Chromosome Location:
          Chr.6: 35468401 - 35470781 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05032352_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPL10A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007104.4 NP_009035.3 6 557 106
          XM_017010900.1 XP_016866389.1 6 812 106
          GenBank mRNA AB082926.1 - 1 538 106
          AF086557.1 - 1 186 106
          AK092386.1 - 1 1210 106
          AK313181.1 - 6 556 106
          BC006791.1 - 6 538 2901249 106
          BC011366.1 - 5 529 2986088 106
          BC070216.1 - 1 545 6495090 106
          BC116456.1 - 1 490 106
          BT020040.1 - 5 523 106
          BT020041.1 - 5 523 106
          U12404.1 - 6 538 106

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RPL10A

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          maturation of LSU-rRNA
          rRNA processing
          translation
          translational initiation
          SRP-dependent cotranslational protein targeting to membrane
          viral transcription
          RNA binding
          structural constituent of ribosome
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000322203.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000464112.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000467020.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000478340.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 ENST00000490335.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 NM_007104.4 Refseq
          Affymetrix Human Clariom D Assay TC0600007800.hg.1 NM_007104.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0