Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000618718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000618718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000618718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
ENST00000618718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000367651.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000536159.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000537332.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000618718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000618718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000618718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
ENST00000618718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_001168389.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013361.hg.1 |
NM_006079.4 |
Refseq |