Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05043600_s1
          See other SETMAR GE Assays ›
          Gene Symbol:
          SETMAR
          Gene Name:
          SET domain and mariner transposase fusion gene
          Gene Aliases:
          METNASE, Mar1
          Chromosome Location:
          Chr.3: 4303304 - 4320649 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05043600_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SETMAR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006713292.3 XP_006713355.1 1 264 92
          XM_006713294.3 XP_006713357.1 1 241 92
          XM_017007032.1 XP_016862521.1 1 242 92
          GenBank mRNA AK311418.1 - 1 212 92
          BG479260.1 - 1 194 91

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_006713294.3, XM_017007032.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA double-strand break processing
          DNA catabolic process, endonucleolytic
          double-strand break repair via nonhomologous end joining
          cell population proliferation
          histone H3-K36 methylation
          DNA integration
          replication fork processing
          mitotic DNA integrity checkpoint signaling
          histone H3-K4 methylation
          nucleic acid phosphodiester bond hydrolysis
          histone H3-K36 dimethylation
          positive regulation of DNA topoisomerase (ATP-hydrolyzing) activity
          positive regulation of double-strand break repair via nonhomologous end joining
          negative regulation of chromosome organization
          single-stranded DNA endodeoxyribonuclease activity
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          endonuclease activity
          protein binding
          zinc ion binding
          histone methyltransferase activity (H3-K4 specific)
          protein homodimerization activity
          DNA topoisomerase binding
          histone methyltransferase activity (H3-K36 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000358950.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000462115.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000462115.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000462115.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000490691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 ENST00000490691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358065.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000358950.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000413809.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000425863.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000430981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000462115.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000462115.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000462115.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000490691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 ENST00000490691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001243723.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_001276325.1 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006471.hg.1 NM_006515.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0