Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000462582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000462582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000462582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000332780.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000459846.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000462582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000462582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000462582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000471811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
ENST00000476495.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007367.hg.1 |
NM_173546.2 |
Refseq |