Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05046945_s1
          See other CDKN1B GE Assays ›
          Gene Symbol:
          CDKN1B
          Gene Name:
          cyclin dependent kinase inhibitor 1B
          Gene Aliases:
          CDKN4, KIP1, MEN1B, MEN4, P27KIP1
          Chromosome Location:
          Chr.12: 12717270 - 12722383 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05046945_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDKN1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004064.4 NP_004055.1 3 1525 116
          GenBank mRNA AK298335.1 - 4 1044 116
          AY004255.1 - 3 1107 116
          BC001971.1 - 3 1332 3458141 116
          CN311141.1 - 2 448 116

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection GE Endogenous Controls Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDKN1B

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          G1/S transition of mitotic cell cycle
          response to hypoxia
          potassium ion transport
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          Notch signaling pathway
          sensory perception of sound
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          response to xenobiotic stimulus
          response to glucose
          positive regulation of cell death
          negative regulation of cell growth
          positive regulation of microtubule polymerization
          response to estradiol
          negative regulation of kinase activity
          negative regulation of phosphorylation
          negative regulation of apoptotic process
          response to amino acid
          response to peptide hormone
          positive regulation of protein catabolic process
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          response to cadmium ion
          autophagic cell death
          inner ear development
          negative regulation of cellular component movement
          regulation of cell cycle
          negative regulation of epithelial cell proliferation involved in prostate gland development
          cellular response to antibiotic
          cellular response to lithium ion
          cellular response to organic cyclic compound
          negative regulation of vascular associated smooth muscle cell proliferation
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          protein binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          protein phosphatase binding
          Hsp70 protein binding
          protein-containing complex binding
          chaperone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000228872.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000396340.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000442489.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 ENST00000477087.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq
          Affymetrix Human Clariom D Assay TC1200006888.hg.1 NM_004064.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0