Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05057817_g1
          See other CKS1B GE Assays ›
          Gene Symbol:
          CKS1B
          Gene Name:
          CDC28 protein kinase regulatory subunit 1B
          Gene Aliases:
          CKS1, PNAS-16, PNAS-18, ckshs1
          Chromosome Location:
          Chr.1: 154974642 - 154979249 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05057817_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CKS1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001826.2 NP_001817.1 2 211 70
          NR_024163.1 2 319 70
          GenBank mRNA AF274941.1 - 1 133 70
          AF274943.1 - 2 139 70
          AF279897.1 - 2 133 70
          AK307066.1 - 2 267 70
          AK311769.1 - 2 172 70
          BC007751.2 - 2 154 4110344 70
          BC015629.2 - 2 133 3162564 70
          BC070319.1 - 2 172 4738841 70
          BC070320.1 - 2 172 6494754 70
          BQ883992.1 - 2 150 70
          BT007196.1 - 2 107 70
          CB161621.1 - 2 270 70
          CN347951.1 - 2 213 70
          X54941.1 - 2 116 70

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CKS1B

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          regulation of mitotic cell cycle
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of transcription, DNA-templated
          cell division
          protein binding
          protein kinase binding
          histone binding
          ubiquitin binding
          cyclin-dependent protein serine/threonine kinase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368436.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368436.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368436.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368436.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000473344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000473344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000473344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000473344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000477676.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NM_001826.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 NR_024163.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000308987.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368436.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368436.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368436.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368436.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000368439.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000471245.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000473344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000473344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000473344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000473344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000474215.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100010100.hg.1 ENST00000474215.5 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline