Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05060852_s1
          See other CLASP1 GE Assays ›
          Gene Symbol:
          CLASP1
          Gene Name:
          cytoplasmic linker associated protein 1
          Gene Aliases:
          MAST1
          Chromosome Location:
          Chr.2: 121337776 - 121649476 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05060852_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CLASP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142273.1 NP_001135745.1 23 2515 100
          NM_001142274.1 NP_001135746.1 22 2491 100
          NM_001207051.1 NP_001193980.1 22 2512 100
          NM_015282.2 NP_056097.1 22 2491 100
          XM_006712377.1 XP_006712440.1 24 2534 100
          XM_006712381.1 XP_006712444.1 22 2483 100
          XM_006712382.1 XP_006712445.1 21 2435 100
          XM_006712383.1 XP_006712446.1 21 2435 100
          XM_006712385.1 XP_006712448.1 16 1534 100
          XM_011510848.1 XP_011509150.1 22 2462 100
          XM_011510849.1 XP_011509151.1 22 2462 100
          XM_017003657.1 XP_016859146.1 24 2534 100
          XM_017003658.1 XP_016859147.1 24 2534 100
          XM_017003659.1 XP_016859148.1 24 2534 100
          XM_017003660.1 XP_016859149.1 24 2534 100
          XM_017003661.1 XP_016859150.1 23 2486 100
          XM_017003662.1 XP_016859151.1 22 2462 100
          XM_017003663.1 XP_016859152.1 22 2462 100
          XM_017003664.1 XP_016859153.1 22 2483 100
          XM_017003665.1 XP_016859154.1 24 2534 100
          XM_017003666.1 XP_016859155.1 24 2534 100
          XM_017003667.1 XP_016859156.1 23 2486 100
          XM_017003668.1 XP_016859157.1 22 2483 100
          XM_017003669.1 XP_016859158.1 22 2462 100
          XM_017003670.1 XP_016859159.1 21 2435 100
          XM_017003671.1 XP_016859160.1 22 2462 100
          XM_017003672.1 XP_016859161.1 22 2483 100
          XM_017003673.1 XP_016859162.1 23 2486 100
          XM_017003674.1 XP_016859163.1 23 2486 100
          XM_017003675.1 XP_016859164.1 22 2462 100
          XM_017003676.1 XP_016859165.1 17 3368 100
          XM_017003677.1 XP_016859166.1 16 3320 100
          XM_017003678.1 XP_016859167.1 15 3295 100
          XM_017003679.1 XP_016859168.1 15 3296 100
          XM_017003680.1 XP_016859169.1 15 3296 100
          XM_017003681.1 XP_016859170.1 16 3319 100
          XM_017003682.1 XP_016859171.1 16 3319 100
          XM_017003683.1 XP_016859172.1 15 3315 100
          XM_017003684.1 XP_016859173.1 16 2301 100
          XM_017003685.1 XP_016859174.1 15 3294 100
          XM_017003686.1 XP_016859175.1 15 3315 100
          XM_017003687.1 XP_016859176.1 16 2301 100
          XM_017003688.1 XP_016859177.1 15 3294 100
          XM_017003689.1 XP_016859178.1 16 2300 100
          XM_017003690.1 XP_016859179.1 24 2534 100
          XM_017003691.1 XP_016859180.1 23 2507 100
          GenBank mRNA AB014522.1 - 14 1354 100
          AF347693.1 - 21 2266 100
          AK074338.1 - 8 747 101
          AK091140.1 - 22 2473 100
          AK297259.1 - 16 1469 100
          AK299331.1 - 16 2019 100
          AK302393.1 - 16 1551 100
          AY425956.1 - 4 226 100
          BC112940.1 - 22 2435 5501463 100
          BC132723.1 - 23 2477 40146745 100
          BC144107.1 - 22 2474 9052627 100
          BX648933.1 - 3 116 100
          CR933722.1 - 15 1443 100

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CLASP1

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          microtubule cytoskeleton organization
          microtubule bundle formation
          vesicle targeting
          microtubule nucleation
          negative regulation of microtubule depolymerization
          Golgi organization
          mitotic spindle organization
          sister chromatid cohesion
          establishment or maintenance of cell polarity
          exit from mitosis
          regulation of gastrulation
          positive regulation of epithelial cell migration
          regulation of epithelial to mesenchymal transition
          astral microtubule organization
          microtubule organizing center organization
          negative regulation of microtubule polymerization or depolymerization
          positive regulation of microtubule polymerization
          microtubule anchoring
          establishment of mitotic spindle localization
          positive regulation of exocytosis
          establishment of spindle orientation
          cell division
          negative regulation of stress fiber assembly
          regulation of focal adhesion assembly
          regulation of microtubule cytoskeleton organization
          positive regulation of extracellular matrix disassembly
          establishment of epithelial cell polarity
          mitotic spindle assembly
          negative regulation of wound healing, spreading of epidermal cells
          positive regulation of basement membrane assembly involved in embryonic body morphogenesis
          dystroglycan binding
          protein binding
          microtubule binding
          kinetochore binding
          microtubule plus-end binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0