Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05276550_g1
          See other LOC100506290 GE Assays ›
          Gene Symbol:
          LOC100506290 FKSG48 FKSG49 FKTN LOC105374363 LOC402469 METTL21A ORC6
          Gene Name:
          uncharacterized LOC100506290
          FKSG48
          FKSG49
          fukutin
          uncharacterized LOC105374363
          uncharacterized LOC402469
          methyltransferase like 21A
          origin recognition complex subunit 6
          Gene Aliases:
          -
          -
          FKSG52, FKSG55, FKSG56, FKSG57, FKSG60, FKSG61, FKSG64, FKSG65, FKSG66, FKSG67, FKSG68, FKSG70
          CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
          -
          FKSG50, FKSG53, FKSG54
          FAM119A, HCA557b, HSPA-KMT
          ORC6L
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.9: 105558117 - 105655950 on Build GRCh38
          Chromosome Location:
          Chr.2: 27356246 - 27358664 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.2: 207580631 - 207625928 on Build GRCh38
          Chromosome Location:
          Chr.16: 46689646 - 46698394 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          Assay ID Hs05276550_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100506290
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF336880.1 - 1 233 75
          Gene Symbol: FKSG48
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF336875.1 - 1 227 75
          Gene Symbol: FKSG49
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF336879.1 - 1 75
          AF336883.1 - 1 75
          AF336887.1 - 1 75
          AF338193.1 - 1 75
          AF338194.1 - 1 75
          AF338195.1 - 1 75
          AF338196.1 - 1 75
          AF338198.1 - 1 75
          AF338199.1 - 1 75
          AF469044.1 - 1 75
          Gene Symbol: FKTN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB609007.1 - 75
          Gene Symbol: LOC105374363
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK127582.1 - 2 75
          Gene Symbol: LOC402469
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF336877.1 - 1 224 75
          AF336881.1 - 1 75
          AF338191.1 - 1 209 75
          Gene Symbol: METTL21A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127395.2 NP_001120867.1 4 2274 75
          NM_145280.5 NP_660323.3 4 2135 75
          Gene Symbol: ORC6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK024077.1 - 4 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK126186.1 - 2 Chr. 17 - 67465453 - 67471240 [+] on Build GRCh38 75
          GenBankmRNA AK095590.1 - 2 Chr. 22 - 17556002 - 17562040 [+] on Build GRCh38 75
          GenBankmRNA BC016696.1 - 2 Chr. 11 - 86109497 - 86112828 [+] on Build GRCh38 75

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100506290
          Click here to find siRNAs for Gene FKSG48
          Click here to find siRNAs for Gene FKSG49
          Click here to find siRNAs for Gene FKTN
          Click here to find siRNAs for Gene LOC105374363
          Click here to find siRNAs for Gene LOC402469
          Click here to find siRNAs for Gene METTL21A
          Click here to find siRNAs for Gene ORC6

          Panther Classification:

          Molecular Function -

          replication origin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein methylation
          peptidyl-lysine methylation
          regulation of ATPase activity
          protein binding
          protein-lysine N-methyltransferase activity
          Hsp70 protein binding
          ATPase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0