Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05287363_g1
          See other CEP170 GE Assays ›
          Gene Symbol:
          CEP170 RAD51B
          Gene Name:
          centrosomal protein 170
          RAD51 paralog B
          Gene Aliases:
          FAM68A, KAB, KIAA0470
          R51H2, RAD51L1, REC2
          Chromosome Location:
          Chr.1: 243124428 - 243255936 on Build GRCh38
          Chromosome Location:
          Chr.14: 67819779 - 68683118 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs05287363_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CEP170
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014812.2 NP_055627.2 10 1893 126
          XM_006711843.3 XP_006711906.1 9 2602 126
          XM_011544334.2 XP_011542636.1 9 2609 126
          XM_011544335.2 XP_011542637.1 9 2022 126
          XM_011544336.2 XP_011542638.1 9 2928 126
          XM_011544337.2 XP_011542639.1 9 2609 126
          XM_011544338.2 XP_011542640.1 9 2609 126
          XM_011544339.2 XP_011542641.1 9 2609 126
          XM_011544340.2 XP_011542642.1 9 2559 126
          XM_011544341.2 XP_011542643.1 10 2373 126
          XM_011544342.2 XP_011542644.1 9 2559 126
          XM_011544343.2 XP_011542645.1 8 2082 126
          XM_017002932.1 XP_016858421.1 9 2522 126
          XM_017002933.1 XP_016858422.1 10 2193 126
          XM_017002934.1 XP_016858423.1 10 1836 126
          XM_017002935.1 XP_016858424.1 10 2373 126
          XM_017002936.1 XP_016858425.1 10 1836 126
          XM_017002937.1 XP_016858426.1 10 2373 126
          XM_017002938.1 XP_016858427.1 10 2373 126
          XM_017002939.1 XP_016858428.1 10 2193 126
          XM_017002940.1 XP_016858429.1 10 1836 126
          XM_017002941.1 XP_016858430.1 10 2193 126
          XM_017002942.1 XP_016858431.1 10 2373 126
          XM_017002943.1 XP_016858432.1 10 2193 126
          XM_017002946.1 XP_016858435.1 9 1896 126
          XM_017002950.1 XP_016858439.1 9 1896 126
          GenBank mRNA AB022657.1 - 9 1538 126
          AK296812.1 - 9 1317 126
          Gene Symbol: RAD51B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017021545.1 XP_016877034.1 8 1046 126

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CEP170
          Click here to find siRNAs for Gene RAD51B

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic DNA recombinase assembly
          double-strand break repair via homologous recombination
          DNA synthesis involved in DNA repair
          strand displacement
          blastocyst growth
          DNA repair
          DNA recombination
          mitotic recombination
          reciprocal meiotic recombination
          blood coagulation
          positive regulation of cell proliferation
          response to ionizing radiation
          positive regulation of G2/M transition of mitotic cell cycle
          strand invasion
          somite development
          protein binding
          recombinase activity
          four-way junction DNA binding
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          endodeoxyribonuclease activity
          ATP binding
          DNA-dependent ATPase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline