Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05300734_g1
          See other BCAR1 GE Assays ›
          Gene Symbol:
          BCAR1
          Gene Name:
          BCAR1, Cas family scaffolding protein
          Gene Aliases:
          CAS, CAS1, CASS1, CRKAS, P130Cas
          Chromosome Location:
          Chr.16: 75229030 - 75268053 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05300734_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BCAR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170714.1 NP_001164185.1 3 744 76
          NM_001170715.1 NP_001164186.1 2 449 76
          NM_001170716.1 NP_001164187.1 2 406 76
          NM_001170717.1 NP_001164188.1 2 468 76
          NM_001170718.1 NP_001164189.1 2 493 76
          NM_001170719.1 NP_001164190.1 2 583 76
          NM_014567.3 NP_055382.2 2 468 76
          XM_005256258.4 XP_005256315.1 2 668 76
          XM_005256259.4 XP_005256316.1 2 3580 76
          XM_005256260.4 XP_005256317.1 2 589 76
          XM_011523475.2 XP_011521777.1 2 617 76
          XM_017023897.1 XP_016879386.1 2 1175 76
          XM_017023898.1 XP_016879387.1 2 1175 76
          XM_017023899.1 XP_016879388.1 2 1175 76
          GenBank mRNA AB040024.1 - 2 463 76
          AF218451.1 - 2 446 76
          AJ242987.2 - 2 443 76
          AK027608.1 - 2 493 76
          AK124526.1 - 2 1888 76
          AK124815.1 - 2 452 76
          AK293808.1 - 3 744 76
          AK294513.1 - 2 499 76
          AK295809.1 - 2 378 76
          AK314722.1 - 2 468 76
          AY545071.1 - 1 322 76
          BC062556.1 - 1 323 6427808 76
          DC317947.1 - 2 409 76

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BCAR1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          actin filament organization
          cell adhesion
          epidermal growth factor receptor signaling pathway
          G-protein coupled receptor signaling pathway
          integrin-mediated signaling pathway
          cell proliferation
          insulin receptor signaling pathway
          positive regulation of endothelial cell migration
          cell migration
          positive regulation of cell migration
          cellular response to hepatocyte growth factor stimulus
          regulation of apoptotic process
          platelet-derived growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          neurotrophin TRK receptor signaling pathway
          hepatocyte growth factor receptor signaling pathway
          antigen receptor-mediated signaling pathway
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          cell division
          cell chemotaxis
          signal transducer activity
          protein binding
          SH3 domain binding
          protein kinase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline