Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs05335722_s1
See other SEPT9 GE Assays ›
Gene Symbol:
SEPT9
Gene Name:
septin 9
Gene Aliases:
AF17q25, MSF, MSF1, NAPB, PNUTL4, SINT1, SeptD1
Chromosome Location:
Chr.17: 77281410 - 77500596 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs05335722_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SEPT9
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001113491.1
NP_001106963.1
12
3409
61
NM_001113492.1
NP_001106964.1
12
3584
61
NM_001113493.1
NP_001106965.1
11
3570
61
NM_001113494.1
NP_001106966.1
11
3523
61
NM_001113495.1
NP_001106967.1
10
3223
61
NM_001113496.1
NP_001106968.1
10
2818
61
NM_001293695.1
NP_001280624.1
11
3352
61
NM_001293696.1
NP_001280625.1
10
2634
61
NM_001293697.1
NP_001280626.1
10
2847
61
NM_001293698.1
NP_001280627.1
10
2933
61
NM_006640.4
NP_006631.2
11
4041
61
XM_005256962.1
XP_005257019.1
10
2691
61
XM_006721643.2
XP_006721706.1
13
3512
61
XM_006721644.1
XP_006721707.1
10
2734
61
XM_011524204.1
XP_011522506.1
13
3609
61
XM_011524206.1
XP_011522508.1
11
3329
61
XM_011524207.1
XP_011522509.1
11
3342
61
XM_011524208.2
XP_011522510.1
10
2600
61
XM_017024031.1
XP_016879520.1
11
2941
61
XM_017024032.1
XP_016879521.1
10
2940
61
GenBank mRNA
AB023208.1
-
11
3524
61
AF142408.1
-
11
3482
61
AF142569.1
-
1
1436
61
AF189712.2
-
13
3539
61
AF189713.3
-
12
3382
61
AJ312319.1
-
11
3571
61
AJ312320.1
-
10
2613
61
AK022493.1
-
6
1498
61
AK056495.1
-
10
3224
61
AL080131.1
-
7
2229
61
BC018877.1
-
1
726
61
BC021192.2
-
11
3265
3346534
61
BC054004.1
-
13
3240
6146448
61
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
3' Most
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene SEPT9
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
protein heterooligomerization
cell division
cell-cell adhesion
positive regulation of nonmotile primary cilium assembly
GTPase activity
protein binding
GTP binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5756d5bff6-86w5v:80/100.66.130.62:80.
git-commit: 27702359083d4c7b0962b032f2611a59cac079b5
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0