Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05562544_g1
          See other CAP1 GE Assays ›
          Gene Symbol:
          CAP1
          Gene Name:
          adenylate cyclase associated protein 1
          Gene Aliases:
          CAP, CAP1-PEN
          Chromosome Location:
          Chr.1: 40040070 - 40072649 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05562544_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CAP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105530.1 NP_001099000.1 12 - 13 1552 108
          NM_006367.3 NP_006358.1 12 - 13 1570 108
          XM_005270368.1 XP_005270425.1 12 - 13 1410 108
          XM_011540509.1 XP_011538811.1 13 - 14 1963 108
          XM_011540510.1 XP_011538812.1 13 - 14 1960 108
          XM_011540511.1 XP_011538813.1 12 - 13 1394 108
          XM_011540512.1 XP_011538814.1 12 - 13 1513 108
          XM_011540513.2 XP_011538815.1 12 - 13 1405 108
          XM_011540514.1 XP_011538816.1 13 - 14 1520 108
          XM_011540515.1 XP_011538817.1 13 - 14 1960 108
          XM_017000067.1 XP_016855556.1 12 - 13 1478 108
          XM_017000068.1 XP_016855557.1 13 - 14 1522 108
          XM_017000069.1 XP_016855558.1 13 - 14 1733 108
          XM_017000070.1 XP_016855559.1 12 - 13 1752 108
          XM_017000071.1 XP_016855560.1 12 - 13 1710 108
          XM_017000072.1 XP_016855561.1 12 - 13 1646 108
          XM_017000073.1 XP_016855562.1 12 - 13 1467 108
          GenBank mRNA AK096193.1 - 12 - 13 1404 108
          AK222513.1 - 12 - 13 1411 108
          AK295396.1 - 13 - 14 1332 108
          AK297829.1 - 12 - 13 1279 108
          AK298094.1 - 12 - 13 1250 108
          AK298110.1 - 12 - 13 1354 108
          AK300861.1 - 11 - 12 1249 108
          AK315731.1 - 12 - 13 1457 108
          BC013963.2 - 12 - 13 1387 2963922 108
          BC020367.1 - 1 - 2 109 108
          BC095440.1 - 12 - 13 1392 30370256 108
          BT007152.1 - 11 - 12 1341 108
          CR457409.1 - 11 - 12 1341 108
          L12168.1 - 11 - 12 1380 108
          M98474.1 - 12 - 13 1411 108

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CAP1

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          ameboidal-type cell migration
          receptor-mediated endocytosis
          establishment or maintenance of cell polarity
          signal transduction
          activation of adenylate cyclase activity
          actin polymerization or depolymerization
          actin binding
          adenylate cyclase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0