Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05635549_g1
          See other LOC102723407 GE Assays ›
          Gene Symbol:
          LOC102723407 SKAP2
          Gene Name:
          putative V-set and immunoglobulin domain-containing-like protein IGHV4OR15-8
          src kinase associated phosphoprotein 2
          Gene Aliases:
          IGH, IGH@, IGHV4, IGHV4-30, IGHV4-b, IGVH, IGVH4-39, IGVH4-b, IGVH4b, IgA, IgT, ighv
          PRAP, RA70, SAPS, SCAP2, SKAP-HOM, SKAP55R
          Chromosome Location:
          Chr.14|NT_187381: 41382 - 42173 on Build GRCh38
          Chromosome Location:
          Chr.7: 26654772 - 26864743 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs05635549_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC102723407
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011546198.1 XP_011544500.1 2 310 117
          GenBank mRNA AF103048.1 - 1 166 117
          AF103313.1 - 1 158 117
          AF174124.1 - 191 117
          AF396470.1 - 1 191 117
          AF396478.1 - 1 191 117
          AF455548.1 - 1 191 117
          AJ244933.1 - 1 191 117
          AJ389176.1 - 1 191 117
          AJ414019.1 - 1 191 117
          AJ579202.1 - 1 194 117
          AY393167.1 - 1 144 117
          AY671339.1 - 1 184 117
          DQ100904.1 - 1 187 117
          DQ454522.1 - 1 113 117
          DQ454670.1 - 1 116 117
          DQ454700.1 - 1 104 117
          DQ454827.1 - 1 113 117
          DQ454834.1 - 1 107 117
          DQ454952.1 - 1 107 117
          DQ454953.1 - 1 110 117
          DQ523999.1 - 1 188 117
          DQ524000.1 - 1 188 117
          EF175423.1 - 126 117
          EF175437.1 - 126 117
          EU099126.1 - 1 140 117
          FJ039783.1 - 1 191 117
          HM152512.1 - 1 191 117
          HM152518.1 - 2 194 117
          HM996243.1 - 1 193 117
          JN376176.1 - 1 116 117
          JN376188.1 - 1 116 117
          JN376230.1 - 1 116 117
          JN376231.1 - 1 116 117
          JN376232.1 - 1 116 117
          JN376233.1 - 1 116 117
          JN376234.1 - 1 116 117
          JN376235.1 - 1 116 117
          JN376309.1 - 1 116 117
          KU674890.1 - 1 144 117
          S45207.1 - 356 117
          S45947.1 - 248 117
          Y14936.1 - 1 170 117
          Z14195.1 - 2 285 117
          Gene Symbol: SKAP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF174049.1 - 1 188 117

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          immunoglobulin

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0