Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05645349_g1
          See other TRIM56 GE Assays ›
          Gene Symbol:
          TRIM56 FKSG49 FKTN LOC105374363 LOC402469 METTL21A NEK5 ORC6 SGK494 TDRKH
          Gene Name:
          tripartite motif containing 56
          FKSG49
          fukutin
          uncharacterized LOC105374363
          uncharacterized LOC402469
          methyltransferase like 21A
          NIMA related kinase 5
          origin recognition complex subunit 6
          uncharacterized serine/threonine-protein kinase SgK494
          tudor and KH domain containing
          Gene Aliases:
          RNF109
          FKSG52, FKSG55, FKSG56, FKSG57, FKSG60, FKSG61, FKSG64, FKSG65, FKSG66, FKSG67, FKSG68, FKSG70
          CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
          -
          FKSG50, FKSG53, FKSG54
          FAM119A, HCA557b, HSPA-KMT
          -
          ORC6L
          -
          TDRD2
          Chromosome Location:
          Chr.7: 101085424 - 101097967 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.9: 105558117 - 105655950 on Build GRCh38
          Chromosome Location:
          Chr.2: 27356246 - 27358664 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.2: 207580631 - 207625928 on Build GRCh38
          Chromosome Location:
          Chr.13: 52064762 - 52129092 on Build GRCh38
          Chromosome Location:
          Chr.16: 46689646 - 46698394 on Build GRCh38
          Chromosome Location:
          Chr.17: 28607964 - 28614193 on Build GRCh38
          Chromosome Location:
          Chr.1: 151769354 - 151790576 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          Assay ID Hs05645349_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRIM56
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011516589.2 XP_011514891.1 2 5932 72
          Gene Symbol: FKSG49
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF336887.1 - 1 72
          AF338196.1 - 1 72
          AF338199.1 - 1 72
          Gene Symbol: FKTN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB609007.1 - 3908 72
          Gene Symbol: LOC105374363
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK127582.1 - 2 2804 72
          Gene Symbol: LOC402469
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF336877.1 - 1 72
          AF336881.1 - 1 72
          AF338191.1 - 1 72
          Gene Symbol: METTL21A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127395.2 NP_001120867.1 4 72
          NM_145280.5 NP_660323.3 4 72
          Gene Symbol: NEK5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK126330.1 - 9 72
          Gene Symbol: ORC6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK024077.1 - 4 72
          Gene Symbol: SGK494
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK294189.1 - 8 1863 72
          Gene Symbol: TDRKH
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK225160.1 - 15 3462 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK126186.1 - 2 3131 Chr. 17 - 67465453 - 67471240 [+] on Build GRCh38 72
          GenBankmRNA BC034305.1 - 2 1333 Chr. 20 - 17778326 - 17780095 [+] on Build GRCh38 72
          GenBankmRNA AK129942.1 - 1 Chr. 11 - 85852557 - 85854943 [-] on Build GRCh38 72
          GenBankmRNA AK056289.1 - 5 1748 Chr. 7 - 68047970 - 68084254 [+] on Build GRCh38 72
          GenBankmRNA BC016696.1 - 2 543 Chr. 11 - 86109497 - 86112828 [+] on Build GRCh38 72

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRIM56
          Click here to find siRNAs for Gene FKSG49
          Click here to find siRNAs for Gene FKTN
          Click here to find siRNAs for Gene LOC105374363
          Click here to find siRNAs for Gene LOC402469
          Click here to find siRNAs for Gene METTL21A
          Click here to find siRNAs for Gene NEK5
          Click here to find siRNAs for Gene ORC6
          Click here to find siRNAs for Gene SGK494
          Click here to find siRNAs for Gene TDRKH

          Panther Classification:

          Molecular Function -

          replication origin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of type I interferon production
          interferon-beta production
          response to type I interferon
          defense response to virus
          protein K63-linked ubiquitination
          protein methylation
          peptidyl-lysine methylation
          regulation of ATPase activity
          ubiquitin-protein transferase activity
          zinc ion binding
          ligase activity
          poly(A) RNA binding
          protein binding
          protein-lysine N-methyltransferase activity
          Hsp70 protein binding
          ATPase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0