Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05659735_s1
          See other MAP4K4 GE Assays ›
          Gene Symbol:
          MAP4K4
          Gene Name:
          mitogen-activated protein kinase kinase kinase kinase 4
          Gene Aliases:
          FLH21957, HEL-S-31, HGK, MEKKK4, NIK
          Chromosome Location:
          Chr.2: 101697703 - 101894690 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05659735_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAP4K4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001242559.1 NP_001229488.1 30 4576 118
          NM_001242560.1 NP_001229489.1 31 4937 118
          NM_004834.4 NP_004825.3 30 4727 118
          NM_145686.3 NP_663719.2 31 5051 118
          NM_145687.3 NP_663720.1 31 4495 118
          XM_005264048.2 XP_005264105.1 33 5080 118
          XM_005264050.3 XP_005264107.1 32 4942 118
          XM_005264052.3 XP_005264109.1 32 4939 118
          XM_005264057.3 XP_005264114.1 32 4873 118
          XM_005264058.3 XP_005264115.1 31 4864 118
          XM_005264059.3 XP_005264116.1 32 4780 118
          XM_005264060.3 XP_005264117.1 31 4768 118
          XM_005264062.3 XP_005264119.1 31 4711 118
          XM_005264063.3 XP_005264120.1 31 4708 118
          XM_005264064.3 XP_005264121.1 30 4702 118
          XM_005264065.3 XP_005264122.1 31 4618 118
          XM_005264066.3 XP_005264123.1 31 4615 118
          XM_005264068.3 XP_005264125.1 30 4609 118
          XM_005264069.2 XP_005264126.1 30 4555 118
          XM_005264071.2 XP_005264128.1 29 4483 118
          XM_006712865.2 XP_006712928.1 32 4849 118
          XM_006712868.2 XP_006712931.1 31 4594 118
          XM_006712869.2 XP_006712932.1 30 4495 118
          XM_017005349.1 XP_016860838.1 33 4984 118
          XM_017005350.1 XP_016860839.1 32 4918 118
          XM_017005351.1 XP_016860840.1 31 4861 118
          XM_017005352.1 XP_016860841.1 32 4849 118
          XM_017005353.1 XP_016860842.1 32 4846 118
          XM_017005354.1 XP_016860843.1 32 4825 118
          XM_017005355.1 XP_016860844.1 31 4816 118
          XM_017005356.1 XP_016860845.1 31 4771 118
          XM_017005357.1 XP_016860846.1 32 4756 118
          XM_017005358.1 XP_016860847.1 32 4753 118
          XM_017005359.1 XP_016860848.1 31 4750 118
          XM_017005360.1 XP_016860849.1 31 4747 118
          XM_017005361.1 XP_016860850.1 31 4726 118
          XM_017005362.1 XP_016860851.1 31 4723 118
          XM_017005363.1 XP_016860852.1 31 4687 118
          XM_017005364.1 XP_016860853.1 30 4675 118
          XM_017005365.1 XP_016860854.1 31 4657 118
          XM_017005366.1 XP_016860855.1 31 4657 118
          XM_017005367.1 XP_016860856.1 31 4630 118
          XM_017005368.1 XP_016860857.1 31 4561 118
          XM_017005369.1 XP_016860858.1 30 4516 118
          XM_017005370.1 XP_016860859.1 30 4492 118
          XM_017005371.1 XP_016860860.1 29 4486 118
          XM_017005372.1 XP_016860861.1 30 4426 118
          XM_017005373.1 XP_016860862.1 29 4417 118
          XM_017005374.1 XP_016860863.1 30 4402 118
          XM_017005375.1 XP_016860864.1 29 4393 118
          GenBank mRNA BC045694.1 - 1 211 118
          BU595561.1 - 1 234 118
          EU794634.1 - 31 4545 118

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAP4K4

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          signal transduction by protein phosphorylation
          microvillus assembly
          intracellular signal transduction
          negative regulation of apoptotic process
          regulation of JNK cascade
          creatine kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0