Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06609250_s1
          See other MESTIT1 GE Assays ›
          Gene Symbol:
          MESTIT1
          Gene Name:
          MEST intronic transcript 1, antisense RNA
          Gene Aliases:
          MEST-AS1, MEST-IT, MEST-IT1, NCRNA00040, PEG1-AS
          Chromosome Location:
          Chr.7: 130487057 - 130491172 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06609250_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MESTIT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004382.1 2 1880 109

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P83803 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3418877 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 32857 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9983 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21515 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3418877 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P83803 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P83803 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3418877 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26039677 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781766 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26039667 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26039678 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26039676 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26039676 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781768 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26039667 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26039677 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781764 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781766 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781762 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05781767 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26039678 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P83803 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781766 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781764 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781778 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781738 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781762 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16018167 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16018151 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781731 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781759 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781721 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781760 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781772 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781737 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781733 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16018153 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781741 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16018164 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781745 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781735 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781717 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781751 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16018156 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16018154 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16018160 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781723 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781747 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781781 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781727 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781749 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781729 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781779 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781722 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781774 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781720 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781769 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781753 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781743 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781755 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781754 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05781758 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11757201_at - -
          Affymetrix Human Genome U219 Array 11757201_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC07001864.hg.1 ENST00000501789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC07001864.hg.1 NR_004382.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC07001864.hg.1 ENST00000501789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC07001864.hg.1 NR_004382.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07028907.hg.1 TC07001864.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07028907.hg.1 ENST00000501789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07028907.hg.1 NR_004382.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07028906.hg.1 TC07001864.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07028906.hg.1 ENST00000501789.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07028906.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700190852.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700190852.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700190852.hg.1 TC0700012603.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 ENST00000562524.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 ENST00000610046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 ENST00000610046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 ENST00000610046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 ENST00000610046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 ENST00000610046.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay TC0700012603.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700190850.hg.1 ENST00000610046.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700190850.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700190850.hg.1 ENST00000610046.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700190850.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700190850.hg.1 TC0700012603.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700190851.hg.1 ENST00000610046.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700190851.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700190851.hg.1 ENST00000610046.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700190851.hg.1 NR_004382.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700190851.hg.1 TC0700012603.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033544 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033544 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3237149 NR_004382.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3237149 NR_004382.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3237149 NR_004382.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3237149 NR_004382.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline