Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs06627451_s1
See other TRDMT1 GE Assays ›
Gene Symbol:
TRDMT1
Gene Name:
tRNA aspartic acid methyltransferase 1
Gene Aliases:
DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1
Chromosome Location:
Chr.10: 17136785 - 17202071 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs06627451_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TRDMT1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001321006.1
NP_001307935.1
5
791
69
NM_001321007.1
NP_001307936.1
8
1164
69
NM_004412.6
NP_004403.1
7
934
69
XM_005252372.4
XP_005252429.1
7
929
69
XM_005252374.4
XP_005252431.1
7
929
69
XM_005252375.4
XP_005252432.1
5
791
69
XM_005252378.3
XP_005252435.1
8
760
69
XM_006717388.3
XP_006717451.1
7
929
69
XM_011519343.2
XP_011517645.1
6
863
69
XM_011519344.2
XP_011517646.1
6
852
69
XM_011519345.2
XP_011517647.1
6
1265
69
XM_017015800.1
XP_016871289.1
5
791
69
XM_017015801.1
XP_016871290.1
5
791
69
XM_017015802.1
XP_016871291.1
7
3148
69
XM_017015803.1
XP_016871292.1
8
762
69
XM_017015804.1
XP_016871293.1
8
600
69
XM_017015805.1
XP_016871294.1
7
797
69
XM_017015806.1
XP_016871295.1
8
760
69
XM_017015807.1
XP_016871296.1
8
781
69
XM_017015808.1
XP_016871297.1
8
595
69
XM_017015809.1
XP_016871298.1
4
714
69
XM_017015810.1
XP_016871299.1
7
694
69
XM_017015811.1
XP_016871300.1
5
660
69
XM_017015812.1
XP_016871301.1
5
454
69
GenBank mRNA
AF012128.1
-
7
504
69
AF045888.1
-
7
503
69
AF329940.1
-
5
371
69
AF329941.1
-
4
305
69
AF329942.1
-
4
300
69
AF329943.1
-
4
294
69
AF329944.1
-
3
228
69
AJ223333.1
-
7
545
69
AK291417.1
-
7
503
69
AK294100.1
-
6
433
69
AK299027.1
-
5
571
69
BC047733.1
-
7
504
6473993
69
BX537961.1
-
8
754
69
CR450316.1
-
7
497
69
HQ637546.1
-
5
354
69
HQ637547.1
-
6
420
69
HQ637548.1
-
5
359
69
HQ637549.1
-
6
431
69
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene TRDMT1
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
response to amphetamine
tRNA modification
tRNA methylation
C-5 methylation of cytosine
RNA binding
DNA (cytosine-5-)-methyltransferase activity
tRNA methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0