Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06627453_s1
          See other TRDMT1 GE Assays ›
          Gene Symbol:
          TRDMT1
          Gene Name:
          tRNA aspartic acid methyltransferase 1
          Gene Aliases:
          DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1
          Chromosome Location:
          Chr.10: 17136785 - 17202071 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06627453_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRDMT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001321006.1 NP_001307935.1 5 789 69
          NM_001321007.1 NP_001307936.1 8 1162 69
          NM_004412.6 NP_004403.1 7 932 69
          XM_005252372.4 XP_005252429.1 7 927 69
          XM_005252374.4 XP_005252431.1 7 927 69
          XM_005252375.4 XP_005252432.1 5 789 69
          XM_005252378.3 XP_005252435.1 8 758 69
          XM_006717388.3 XP_006717451.1 7 927 69
          XM_011519343.2 XP_011517645.1 6 861 69
          XM_011519344.2 XP_011517646.1 6 850 69
          XM_011519345.2 XP_011517647.1 6 1263 69
          XM_017015800.1 XP_016871289.1 5 789 69
          XM_017015801.1 XP_016871290.1 5 789 69
          XM_017015802.1 XP_016871291.1 7 3146 69
          XM_017015803.1 XP_016871292.1 8 760 69
          XM_017015804.1 XP_016871293.1 8 598 69
          XM_017015805.1 XP_016871294.1 7 795 69
          XM_017015806.1 XP_016871295.1 8 758 69
          XM_017015807.1 XP_016871296.1 8 779 69
          XM_017015808.1 XP_016871297.1 8 593 69
          XM_017015809.1 XP_016871298.1 4 712 69
          XM_017015810.1 XP_016871299.1 7 692 69
          XM_017015811.1 XP_016871300.1 5 658 69
          XM_017015812.1 XP_016871301.1 5 452 69
          GenBank mRNA AF012128.1 - 7 502 69
          AF045888.1 - 7 501 69
          AF329940.1 - 5 369 69
          AF329941.1 - 4 303 69
          AF329942.1 - 4 298 69
          AF329943.1 - 4 292 69
          AF329944.1 - 3 226 69
          AJ223333.1 - 7 543 69
          AK291417.1 - 7 501 69
          AK294100.1 - 6 431 69
          AK299027.1 - 5 569 69
          BC047733.1 - 7 502 6473993 69
          BX537961.1 - 8 752 69
          CR450316.1 - 7 495 69
          HQ637546.1 - 5 352 69
          HQ637547.1 - 6 418 69
          HQ637548.1 - 5 357 69
          HQ637549.1 - 6 429 69

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRDMT1

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          tRNA modification
          tRNA methylation
          C-5 methylation of cytosine
          RNA binding
          DNA (cytosine-5-)-methyltransferase activity
          tRNA methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0