Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06627736_s1
          See other STK11 GE Assays ›
          Gene Symbol:
          STK11
          Gene Name:
          serine/threonine kinase 11
          Gene Aliases:
          LKB1, PJS, hLKB1
          Chromosome Location:
          Chr.19: 1205799 - 1228435 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06627736_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: STK11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000455.4 NP_000446.1 5 1781 66
          XM_005259617.2 XP_005259674.1 5 1291 66
          XM_005259618.3 XP_005259675.1 5 1291 66
          XM_011528209.1 XP_011526511.1 6 1422 66
          GenBank mRNA AB209553.1 - 5 756 66
          AF035625.1 - 5 1004 66
          AF217978.1 - 6 1627 66
          AJ844634.2 - 2 197 66
          AK314858.1 - 5 792 66
          BC007981.2 - 5 1116 3689780 66
          BC019334.1 - 5 937 3946005 66
          EF644172.1 - 5 750 66
          KU178339.1 - 5 666 66
          U63333.1 - 5 666 66

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene STK11

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          tissue homeostasis
          vasculature development
          protein phosphorylation
          autophagy
          cellular response to DNA damage stimulus
          cell cycle arrest
          spermatid development
          axonogenesis
          negative regulation of cell proliferation
          response to ionizing radiation
          positive regulation of autophagy
          establishment of cell polarity
          regulation of Wnt signaling pathway
          negative regulation of cell growth
          positive regulation of transforming growth factor beta receptor signaling pathway
          activation of protein kinase activity
          TCR signalosome assembly
          glucose homeostasis
          anoikis
          positive thymic T cell selection
          protein autophosphorylation
          regulation of dendrite morphogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of axonogenesis
          T cell receptor signaling pathway
          negative regulation of lipid biosynthetic process
          Golgi localization
          regulation of protein kinase B signaling
          canonical Wnt signaling pathway
          negative regulation of epithelial cell proliferation involved in prostate gland development
          cellular response to UV-B
          positive regulation of protein serine/threonine kinase activity
          intrinsic apoptotic signaling pathway by p53 class mediator
          dendrite extension
          positive regulation of protein localization to nucleus
          regulation of signal transduction by p53 class mediator
          negative regulation of TORC1 signaling
          magnesium ion binding
          p53 binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          LRR domain binding
          protein kinase activator activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0