Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06631836_sH
          See other SCN5A GE Assays ›
          Gene Symbol:
          SCN5A
          Gene Name:
          sodium voltage-gated channel alpha subunit 5
          Gene Aliases:
          CDCD2, CMD1E, CMPD2, HB1, HB2, HBBD, HH1, ICCD, IVF, LQT3, Nav1.5, PFHB1, SSS1, VF1
          Chromosome Location:
          Chr.3: 38548061 - 38649673 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06631836_sH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SCN5A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000335.4 NP_000326.2 7 939 85
          NM_001099404.1 NP_001092874.1 7 939 85
          NM_001099405.1 NP_001092875.1 7 939 85
          NM_001160160.1 NP_001153632.1 7 940 85
          NM_001160161.1 NP_001153633.1 7 940 85
          NM_198056.2 NP_932173.1 7 939 85
          XM_011533991.2 XP_011532293.1 7 934 85
          XM_017007017.1 XP_016862506.1 7 934 85
          GenBank mRNA AB158469.2 - 6 787 85
          AB158470.2 - 6 787 85
          AF482988.1 - 6 754 85
          AJ310886.1 - 4 306 85
          AJ310887.1 - 4 306 85
          AJ310888.1 - 4 306 85
          AJ310889.1 - 4 306 85
          AJ310890.1 - 4 306 85
          AJ310891.1 - 4 306 85
          AJ310892.1 - 4 306 85
          AJ310893.1 - 3 177 85
          AJ310894.1 - 4 304 85
          AJ310895.1 - 4 305 85
          AY038064.1 - 6 803 85
          AY148488.1 - 6 745 85
          BC140813.1 - 6 770 9021681 85
          BC144621.1 - 6 770 9053152 85
          EF629346.1 - 6 787 85
          EF629347.1 - 6 787 85
          KC858891.1 - 6 745 85
          M77235.1 - 7 895 85

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SCN5A

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          cardiac ventricle development
          brainstem development
          sodium ion transport
          positive regulation of sodium ion transport
          response to denervation involved in regulation of muscle adaptation
          neuronal action potential
          telencephalon development
          cerebellum development
          sodium ion transmembrane transport
          odontogenesis of dentin-containing tooth
          positive regulation of action potential
          positive regulation of epithelial cell proliferation
          membrane depolarization
          cardiac muscle contraction
          regulation of postsynaptic membrane potential
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of atrial cardiac muscle cell membrane depolarization
          regulation of atrial cardiac muscle cell membrane repolarization
          regulation of ventricular cardiac muscle cell membrane depolarization
          cardiac conduction
          cellular response to calcium ion
          cardiac muscle cell action potential involved in contraction
          regulation of cardiac muscle cell contraction
          ventricular cardiac muscle cell action potential
          membrane depolarization during action potential
          membrane depolarization during cardiac muscle cell action potential
          SA node cell action potential
          AV node cell action potential
          bundle of His cell action potential
          membrane depolarization during AV node cell action potential
          membrane depolarization during SA node cell action potential
          membrane depolarization during Purkinje myocyte cell action potential
          membrane depolarization during bundle of His cell action potential
          AV node cell to bundle of His cell communication
          regulation of heart rate by cardiac conduction
          regulation of sodium ion transmembrane transport
          voltage-gated sodium channel activity
          protein binding
          calmodulin binding
          fibroblast growth factor binding
          enzyme binding
          protein kinase binding
          protein domain specific binding
          ankyrin binding
          ubiquitin protein ligase binding
          ion channel binding
          nitric-oxide synthase binding
          voltage-gated sodium channel activity involved in cardiac muscle cell action potential
          voltage-gated sodium channel activity involved in AV node cell action potential
          voltage-gated sodium channel activity involved in bundle of His cell action potential
          voltage-gated sodium channel activity involved in Purkinje myocyte action potential
          voltage-gated sodium channel activity involved in SA node cell action potential
          scaffold protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0