Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06633147_s1
          See other ATM GE Assays ›
          Gene Symbol:
          ATM
          Gene Name:
          ATM serine/threonine kinase
          Gene Aliases:
          AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
          Chromosome Location:
          Chr.11: 108222484 - 108369102 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06633147_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ATM
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000051.3 NP_000042.3 13 2432 62
          XM_005271561.4 XP_005271618.2 14 2493 62
          XM_005271562.4 XP_005271619.2 13 2566 62
          XM_006718843.3 XP_006718906.1 13 2199 62
          XM_011542840.2 XP_011541142.1 14 2287 62
          XM_011542842.2 XP_011541144.1 12 2241 62
          XM_011542843.2 XP_011541145.1 13 2780 62
          XM_011542844.2 XP_011541146.1 13 2382 62
          XM_011542845.1 XP_011541147.1 5 895 62
          XM_017017789.1 XP_016873278.1 14 2990 62
          XM_017017790.1 XP_016873279.1 13 2387 62
          XM_017017791.1 XP_016873280.1 13 2780 62
          XM_017017792.1 XP_016873281.1 13 2780 62
          GenBank mRNA AF035326.1 - 2 226 62
          AF035327.1 - 2 202 62
          HM437232.1 - 1 152 62
          U33841.1 - 14 2236 62
          X91196.1 - 13 2780 62

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ATM

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          DNA double-strand break processing
          DNA synthesis involved in DNA repair
          strand displacement
          response to hypoxia
          somitogenesis
          pre-B cell allelic exclusion
          DNA replication
          DNA repair
          double-strand break repair via nonhomologous end joining
          protein phosphorylation
          cellular response to DNA damage stimulus
          DNA damage induced protein phosphorylation
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          telomere maintenance via telomerase
          cell cycle arrest
          mitotic spindle assembly checkpoint
          reciprocal meiotic recombination
          signal transduction
          brain development
          heart development
          determination of adult lifespan
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to ionizing radiation
          regulation of autophagy
          histone phosphorylation
          peptidyl-serine phosphorylation
          negative regulation of B cell proliferation
          positive regulation of telomere maintenance via telomerase
          positive regulation of histone phosphorylation
          V(D)J recombination
          phosphatidylinositol-3-phosphate biosynthetic process
          peptidyl-serine autophosphorylation
          lipoprotein catabolic process
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of DNA damage response, signal transduction by p53 class mediator
          positive regulation of neuron apoptotic process
          meiotic telomere clustering
          protein autophosphorylation
          oocyte development
          neuron apoptotic process
          regulation of telomerase activity
          histone mRNA catabolic process
          cellular response to gamma radiation
          cellular response to X-ray
          cellular response to nitrosative stress
          signal transduction involved in mitotic G2 DNA damage checkpoint
          replicative senescence
          establishment of RNA localization to telomere
          establishment of macromolecular complex localization to telomere
          regulation of cellular response to heat
          regulation of signal transduction by p53 class mediator
          negative regulation of TORC1 signaling
          negative regulation of telomere capping
          positive regulation of telomere maintenance via telomere lengthening
          positive regulation of telomerase catalytic core complex assembly
          DNA binding
          protein serine/threonine kinase activity
          DNA-dependent protein kinase activity
          protein binding
          ATP binding
          1-phosphatidylinositol-3-kinase activity
          protein complex binding
          protein dimerization activity
          protein N-terminus binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0