Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06636994_s1
          See other FAS GE Assays ›
          Gene Symbol:
          FAS
          Gene Name:
          Fas cell surface death receptor
          Gene Aliases:
          ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
          Chromosome Location:
          Chr.10: 88968429 - 89017061 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06636994_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FAS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000043.5 NP_000034.1 2 467 70
          NM_001320619.1 NP_001307548.1 2 467 70
          NM_152871.3 NP_690610.1 2 467 70
          NM_152872.3 NP_690611.1 2 467 70
          NR_028033.3 2 467 70
          NR_028034.3 2 467 70
          NR_028035.3 2 467 70
          NR_028036.3 2 467 70
          NR_135313.1 2 467 70
          NR_135314.1 2 458 70
          NR_135315.1 2 458 70
          XM_006717819.3 XP_006717882.1 3 5149 70
          XM_011539764.2 XP_011538066.1 3 538 70
          XM_011539765.2 XP_011538067.1 3 326 70
          XM_011539766.2 XP_011538068.1 3 278 70
          XM_011539767.2 XP_011538069.1 2 201 70
          GenBank mRNA AB209361.1 - 3 3784 70
          AK290978.1 - 2 395 70
          AK311164.1 - 2 315 70
          AK311424.1 - 2 458 70
          AK311499.1 - 2 315 70
          AV715411.1 - 2 303 70
          AY495076.2 - 2 343 70
          BC012479.1 - 2 459 4514272 70
          CR450307.1 - 2 149 70
          DA040126.1 - 2 326 70
          DA699562.1 - 2 467 70
          EU481974.2 - 2 343 70
          FJ200481.1 - 2 149 70
          FJ373045.1 - 2 149 70
          FJ373046.1 - 2 149 70
          FM246457.1 - 2 149 70
          FM246458.1 - 2 149 70
          FM246459.1 - 2 149 70
          M67454.1 - 2 343 70
          X63717.1 - 2 369 70
          X83490.1 - 2 174 70
          X83491.1 - 2 174 70
          X83492.1 - 2 174 70
          X83493.1 - 2 174 70
          X89101.1 - 2 149 70
          Z47993.1 - 2 149 70
          Z47994.1 - 2 149 70
          Z47995.1 - 2 149 70
          Z66556.1 - 2 149 70
          Z70519.1 - 2 149 70
          Z70520.1 - 2 149 70

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FAS

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle atresia
          immunoglobulin production
          renal system process
          protein complex assembly
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          activation-induced cell death of T cells
          inflammatory cell apoptotic process
          inflammatory response
          immune response
          signal transduction
          spermatogenesis
          brain development
          aging
          circadian rhythm
          extrinsic apoptotic signaling pathway via death domain receptors
          response to toxic substance
          gene expression
          regulation of gene expression
          B cell mediated immunity
          telencephalon development
          dendrite regeneration
          positive regulation of protein homooligomerization
          response to lipopolysaccharide
          tumor necrosis factor-mediated signaling pathway
          regulation of cell proliferation
          response to drug
          ovulation cycle
          regulation of apoptotic process
          chordate embryonic development
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAPK cascade
          response to peptide hormone
          negative thymic T cell selection
          regulation of lymphocyte differentiation
          regulation of myeloid cell differentiation
          response to cycloheximide
          spleen development
          negative regulation of B cell activation
          protein homooligomerization
          response to glucocorticoid
          maternal process involved in female pregnancy
          positive regulation of lymphocyte apoptotic process
          cellular response to hydrogen peroxide
          response to growth factor
          cellular response to phenylalanine
          cellular response to mechanical stimulus
          cellular response to cobalt ion
          cellular response to lithium ion
          cellular response to glucose stimulus
          cellular response to interleukin-1
          cellular response to estrogen stimulus
          cellular response to hyperoxia
          cellular response to hypoxia
          cellular response to hydrostatic pressure
          motor neuron apoptotic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          hepatocyte apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          liver regeneration
          necroptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          response to fluoride
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protease binding
          signal transducer activity
          receptor activity
          tumor necrosis factor-activated receptor activity
          protein binding
          kinase binding
          protein complex binding
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0