Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06637224_s1
          See other SERPINA1 GE Assays ›
          Gene Symbol:
          SERPINA1
          Gene Name:
          serpin family A member 1
          Gene Aliases:
          A1A, A1AT, AAT, PI, PI1, PRO2275, alpha1AT
          Chromosome Location:
          Chr.14: 94376747 - 94390692 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06637224_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SERPINA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000295.4 NP_000286.3 1 219 73
          NM_001002236.2 NP_001002236.1 3 512 73
          NM_001127701.1 NP_001121173.1 3 531 73
          NM_001127702.1 NP_001121174.1 2 339 73
          NM_001127703.1 NP_001121175.1 3 494 73
          NM_001127704.1 NP_001121176.1 3 491 73
          NM_001127705.1 NP_001121177.1 3 509 73
          NM_001127706.1 NP_001121178.1 2 302 73
          NM_001127707.1 NP_001121179.1 2 299 73
          XM_017021370.1 XP_016876859.1 2 261 73
          GenBank mRNA AK315637.1 - 2 255 73
          BC011991.1 - 1 190 3859644 73
          BM844817.1 - 1 217 73
          BP296710.1 - 2 242 73
          BP300943.1 - 2 260 73
          BP301274.1 - 2 261 73
          BX247968.1 - 3 439 73
          BX477352.1 - 1 210 73
          CD685665.1 - 3 379 73
          DA428299.1 - 3 497 73
          DA433058.1 - 3 434 73
          DA672472.1 - 3 474 73
          DA675981.1 - 3 451 73
          DA966896.1 - 1 219 73
          X05826.1 - 3 479 73
          X05827.1 - 2 270 73

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK315637.1, BC011991.1, BX247968.1, NM_000295.4, NM_001002236.2, NM_001127701.1, NM_001127702.1, NM_001127703.1, NM_001127704.1, NM_001127705.1, NM_001127706.1, NM_001127707.1, XM_017021370.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SERPINA1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          ER to Golgi vesicle-mediated transport
          acute-phase response
          blood coagulation
          negative regulation of endopeptidase activity
          COPII vesicle coating
          glycoprotein binding
          protease binding
          serine-type endopeptidase inhibitor activity
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix