Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06638279_s1
          See other RBM26 GE Assays ›
          Gene Symbol:
          RBM26
          Gene Name:
          RNA binding motif protein 26
          Gene Aliases:
          ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
          Chromosome Location:
          Chr.13: 79311827 - 79406265 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06638279_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RBM26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286631.1 NP_001273560.1 7 1523 70
          NM_001286632.1 NP_001273561.1 7 1523 70
          NM_022118.4 NP_071401.3 7 1523 70
          XM_005266491.4 XP_005266548.1 7 1560 70
          XM_005266497.1 XP_005266554.1 7 1560 70
          XM_006719857.1 XP_006719920.1 7 1560 70
          XM_011535179.1 XP_011533481.1 7 1560 70
          XM_011535180.2 XP_011533482.1 7 1560 70
          XM_011535181.1 XP_011533483.1 7 1560 70
          XM_011535182.1 XP_011533484.1 7 1560 70
          XM_011535183.2 XP_011533485.1 7 1560 70
          XM_011535184.1 XP_011533486.1 7 1560 70
          XM_011535185.2 XP_011533487.1 7 1560 70
          XM_011535186.2 XP_011533488.1 7 1567 70
          XM_011535187.2 XP_011533489.1 7 1560 70
          XM_011535188.2 XP_011533490.1 7 1567 70
          XM_011535189.1 XP_011533491.1 7 1560 70
          XM_011535190.1 XP_011533492.1 7 1560 70
          XM_011535191.2 XP_011533493.1 7 1560 70
          XM_011535192.2 XP_011533494.1 7 1567 70
          XM_011535193.2 XP_011533495.1 7 1560 70
          XM_011535195.1 XP_011533497.1 7 1258 70
          XM_017020688.1 XP_016876177.1 7 1560 70
          XM_017020689.1 XP_016876178.1 7 1560 70
          XM_017020690.1 XP_016876179.1 7 1567 70
          XM_017020691.1 XP_016876180.1 7 1567 70
          XM_017020692.1 XP_016876181.1 7 1567 70
          XM_017020693.1 XP_016876182.1 7 1560 70
          XM_017020694.1 XP_016876183.1 7 1560 70
          XM_017020695.1 XP_016876184.1 7 1560 70
          XM_017020696.1 XP_016876185.1 7 1567 70
          XM_017020697.1 XP_016876186.1 7 1567 70
          XM_017020698.1 XP_016876187.1 7 1560 70
          XM_017020699.1 XP_016876188.1 7 1567 70
          XM_017020700.1 XP_016876189.1 7 1567 70
          XM_017020701.1 XP_016876190.1 7 1560 70
          XM_017020702.1 XP_016876191.1 7 1560 70
          XM_017020703.1 XP_016876192.1 7 1560 70
          GenBank mRNA AK027339.1 - 2 454 70
          AK027456.1 - 1 172 70
          AK302871.1 - 7 1132 70
          AL834343.1 - 2 436 70
          AX775885.1 - 1 172 70
          BC041655.1 - 7 1090 4384399 70
          BC111697.1 - 7 1084 40038080 70
          BC111739.1 - 7 1084 40038082 70
          BX648672.1 - 7 1081 70

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK027339.1, BC111739.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RBM26

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          negative regulation of phosphatase activity
          nucleotide binding
          protein binding
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix