Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07287128_m1
          See other PRDX3 GE Assays ›
          Gene Symbol:
          PRDX3
          Gene Name:
          peroxiredoxin 3
          Gene Aliases:
          AOP-1, AOP1, HBC189, MER5, PRO1748, SP-22, prx-III
          Chromosome Location:
          Chr.10: 119167699 - 119178865 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07287128_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRDX3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001302272.1 NP_001289201.1 4 - 5 524 74
          NM_006793.4 NP_006784.1 4 - 5 524 74
          NR_126102.1 3 - 4 391 74
          NR_126103.1 2 - 3 249 74
          NR_126105.1 3 - 4 241 74
          GenBank mRNA AK130897.1 - 4 - 5 471 74
          AK222659.1 - 4 - 5 454 74
          AK313169.1 - 4 - 5 462 74
          AV718278.1 - 4 - 5 421 74
          AV761690.1 - 3 - 4 337 74
          BC002685.1 - 4 - 5 455 3607558 74
          BC007062.1 - 4 - 5 479 3960644 74
          BC008435.1 - 4 - 5 463 4081846 74
          BC009601.1 - 4 - 5 456 4134666 74
          BC022373.1 - 4 - 5 469 4775243 74
          BC059169.1 - 4 - 5 456 4133576 74
          BC111397.1 - 4 - 5 463 4246143 74
          BF209381.1 - 3 - 4 180 74
          BG282768.1 - 4 - 5 438 74
          BT020007.1 - 4 - 5 449 74
          BX382389.2 - 4 - 5 473 74
          CD687044.1 - 4 - 5 495 74
          CR450344.1 - 4 - 5 449 74
          D49396.1 - 4 - 5 455 74

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRDX3

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          maternal placenta development
          apoptotic process
          response to oxidative stress
          mitochondrion organization
          positive regulation of cell population proliferation
          peptidyl-cysteine oxidation
          myeloid cell differentiation
          response to lipopolysaccharide
          negative regulation of kinase activity
          cellular response to oxidative stress
          cellular response to reactive oxygen species
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          cell redox homeostasis
          positive regulation of NF-kappaB transcription factor activity
          regulation of mitochondrial membrane potential
          cellular oxidant detoxification
          protein binding
          protein C-terminus binding
          thioredoxin peroxidase activity
          alkyl hydroperoxide reductase activity
          kinase binding
          protein kinase binding
          identical protein binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0