Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs07287184_m1
See other HEXIM2 GE Assays ›
Gene Symbol:
HEXIM2
Gene Name:
hexamethylene bisacetamide inducible 2
Gene Aliases:
L3
Chromosome Location:
Chr.17: 45160174 - 45170040 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs07287184_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: HEXIM2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001303436.1
NP_001290365.1
2 - 3
883
110
NM_001303437.1
NP_001290366.1
3 - 4
1336
110
NM_001303438.1
NP_001290367.1
2 - 3
1188
110
NM_001303439.1
NP_001290368.1
2 - 3
1340
110
NM_001303440.1
NP_001290369.1
2 - 3
300
110
NM_001303441.1
NP_001290370.1
3 - 4
448
110
NM_001303442.1
NP_001290371.1
2 - 3
194
110
NM_001303443.1
NP_001290372.1
3 - 4
342
110
NM_001303444.1
NP_001290373.1
2 - 3
197
110
NM_144608.2
NP_653209.1
3 - 4
1031
110
XM_006721687.3
XP_006721750.1
2 - 3
880
110
XM_011524306.2
XP_011522608.1
3 - 4
352
110
XM_011524307.2
XP_011522609.2
3 - 4
787
110
XM_011524308.2
XP_011522610.2
3 - 4
790
110
XM_017024167.1
XP_016879656.1
3 - 4
465
110
XM_017024168.1
XP_016879657.1
4 - 5
1226
110
XM_017024169.1
XP_016879658.1
3 - 4
1078
110
XM_017024170.1
XP_016879659.1
3 - 4
365
110
GenBank mRNA
AK056946.1
-
3 - 4
308
110
AY598322.1
-
1 - 2
69
110
BC003531.1
-
2 - 3
228
3609101
110
BC012474.1
-
1 - 2
121
4475599
110
BC025970.1
-
2 - 3
158
4559410
110
BG696988.1
-
2 - 3
139
110
BI829228.1
-
2 - 3
422
110
BI829844.1
-
3 - 4
445
110
BP209696.1
-
2 - 3
284
110
BU501076.1
-
2 - 3
327
110
CN425800.1
-
3 - 4
344
110
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
3' Most
Mammalian Gene Collection
Recommended Assay
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene HEXIM2
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
negative regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of transcription, DNA-templated
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding
snRNA binding
7SK snRNA binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0