Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07287866_g1
          See other UBA52 GE Assays ›
          Gene Symbol:
          UBA52
          Gene Name:
          ubiquitin A-52 residue ribosomal protein fusion product 1
          Gene Aliases:
          CEP52, HUBCEP52, L40, RPL40
          Chromosome Location:
          Chr.19: 18563766 - 18577460 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07287866_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UBA52
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001321018.1 NP_001307947.1 1 - 2 735 73
          XM_006722871.2 XP_006722934.1 1 - 2 739 73
          GenBank mRNA CD242660.1 - 1 - 2 314 73

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001321018.1, XM_006722871.2 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          negative regulation of transcription by RNA polymerase II
          MAPK cascade
          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          protein polyubiquitination
          nucleotide-excision repair, DNA damage recognition
          nucleotide-excision repair, DNA duplex unwinding
          stimulatory C-type lectin receptor signaling pathway
          positive regulation of defense response to virus by host
          MyD88-dependent toll-like receptor signaling pathway
          MyD88-independent toll-like receptor signaling pathway
          glycogen biosynthetic process
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair, preincision complex assembly
          nucleotide-excision repair, DNA incision, 5'-to lesion
          nucleotide-excision repair, DNA gap filling
          rRNA processing
          translational initiation
          cellular protein modification process
          ubiquitin-dependent protein catabolic process
          SRP-dependent cotranslational protein targeting to membrane
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          Notch signaling pathway
          I-kappaB kinase/NF-kappaB signaling
          JNK cascade
          fibroblast growth factor receptor signaling pathway
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of necrotic cell death
          Wnt signaling pathway
          endosomal transport
          macroautophagy
          viral life cycle
          virion assembly
          viral transcription
          translesion synthesis
          negative regulation of transforming growth factor beta receptor signaling pathway
          anaphase-promoting complex-dependent catabolic process
          regulation of type I interferon production
          negative regulation of type I interferon production
          tumor necrosis factor-mediated signaling pathway
          nucleotide-excision repair, DNA incision
          ion transmembrane transport
          TRIF-dependent toll-like receptor signaling pathway
          interstrand cross-link repair
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          ERBB2 signaling pathway
          negative regulation of epidermal growth factor receptor signaling pathway
          error-prone translesion synthesis
          DNA damage response, detection of DNA damage
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of mRNA stability
          cellular protein metabolic process
          innate immune response
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of transcription by RNA polymerase II
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          stress-activated MAPK cascade
          Wnt signaling pathway, planar cell polarity pathway
          regulation of transcription from RNA polymerase II promoter in response to hypoxia
          nucleotide-binding oligomerization domain containing signaling pathway
          global genome nucleotide-excision repair
          error-free translesion synthesis
          intracellular transport of virus
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          xenophagy
          regulation of signal transduction by p53 class mediator
          structural constituent of ribosome
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0