Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07288000_g1
          See other CTNNA1 GE Assays ›
          Gene Symbol:
          CTNNA1
          Gene Name:
          catenin alpha 1
          Gene Aliases:
          CAP102, MDPT2
          Chromosome Location:
          Chr.5: 138753386 - 138935034 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07288000_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTNNA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290307.2 NP_001277236.1 11 - 12 1679 93
          NM_001290309.2 NP_001277238.1 10 - 11 1483 93
          NM_001290310.2 NP_001277239.1 11 - 12 1516 93
          NM_001290312.1 NP_001277241.1 5 - 6 999 93
          NM_001323982.1 NP_001310911.1 12 - 13 2149 93
          NM_001323983.1 NP_001310912.1 11 - 12 1831 93
          NM_001323984.1 NP_001310913.1 12 - 13 1697 93
          NM_001323985.1 NP_001310914.1 11 - 12 1679 93
          NM_001323986.1 NP_001310915.1 10 - 11 1586 93
          NM_001323987.1 NP_001310916.1 5 - 6 688 93
          NM_001323988.1 NP_001310917.1 8 - 9 936 93
          NM_001323989.1 NP_001310918.1 6 - 7 784 93
          NM_001323990.1 NP_001310919.1 7 - 8 1148 93
          NM_001323991.1 NP_001310920.1 6 - 7 1095 93
          NM_001323992.1 NP_001310921.1 8 - 9 928 93
          NM_001323993.1 NP_001310922.1 8 - 9 1242 93
          NM_001323994.1 NP_001310923.1 5 - 6 697 93
          NM_001323995.1 NP_001310924.1 8 - 9 912 93
          NM_001323996.1 NP_001310925.1 8 - 9 1247 93
          NM_001323997.1 NP_001310926.1 8 - 9 931 93
          NM_001323998.1 NP_001310927.1 8 - 9 1223 93
          NM_001323999.1 NP_001310928.1 8 - 9 1220 93
          NM_001324000.1 NP_001310929.1 6 - 7 1092 93
          NM_001324001.1 NP_001310930.1 5 - 6 999 93
          NM_001324002.1 NP_001310931.1 5 - 6 999 93
          NM_001324003.1 NP_001310932.1 7 - 8 1148 93
          NM_001324004.1 NP_001310933.1 5 - 6 688 93
          NM_001324005.1 NP_001310934.1 8 - 9 1223 93
          NM_001324006.1 NP_001310935.1 9 - 10 970 93
          NM_001324007.1 NP_001310936.1 8 - 9 1206 93
          NM_001324008.1 NP_001310937.1 6 - 7 746 93
          NM_001324009.1 NP_001310938.1 6 - 7 1057 93
          NM_001324010.1 NP_001310939.1 2 - 3 361 93
          NM_001324011.1 NP_001310940.1 7 - 8 819 93
          NM_001324012.1 NP_001310941.1 3 - 4 454 93
          NM_001324013.1 NP_001310942.1 6 - 7 1028 93
          NM_001903.4 NP_001894.2 11 - 12 1679 93
          GenBank mRNA AK094151.1 - 8 - 9 888 93
          AK296708.1 - 11 - 12 1487 93
          AK300439.1 - 10 - 11 1450 93
          AK304267.1 - 11 - 12 1574 93
          BC000385.2 - 11 - 12 1617 2821111 93
          BC031262.1 - 5 - 6 578 5274351 93
          D13866.1 - 11 - 12 1618 93
          D14705.1 - 11 - 12 1643 93
          HQ589335.1 - 5 - 6 833 93
          L23805.1 - 10 - 11 1559 93
          U03100.1 - 10 - 11 1559 93

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CTNNA1

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          actin filament organization
          cell adhesion
          establishment or maintenance of cell polarity
          negative regulation of neuroblast proliferation
          aging
          male gonad development
          gap junction assembly
          axon regeneration
          adherens junction organization
          cellular protein localization
          odontogenesis of dentin-containing tooth
          apical junction assembly
          response to estrogen
          positive regulation of smoothened signaling pathway
          positive regulation of muscle cell differentiation
          protein heterooligomerization
          cellular response to indole-3-methanol
          epithelial cell-cell adhesion
          negative regulation of cell motility
          negative regulation of integrin-mediated signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          structural molecule activity
          protein binding
          beta-catenin binding
          vinculin binding
          poly(A) RNA binding
          gamma-catenin binding
          cadherin binding
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0