Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs07288001_g1
See other CTNNA1 GE Assays ›
Gene Symbol:
CTNNA1
Gene Name:
catenin alpha 1
Gene Aliases:
CAP102, MDPT2
Chromosome Location:
Chr.5: 138753386 - 138935034 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs07288001_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CTNNA1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001290307.2
NP_001277236.1
13 - 14
2028
99
NM_001290309.2
NP_001277238.1
12 - 13
1832
99
NM_001290310.2
NP_001277239.1
13 - 14
1865
99
NM_001290312.1
NP_001277241.1
7 - 8
1348
99
NM_001323982.1
NP_001310911.1
14 - 15
2498
99
NM_001323983.1
NP_001310912.1
13 - 14
2180
99
NM_001323984.1
NP_001310913.1
14 - 15
2046
99
NM_001323985.1
NP_001310914.1
13 - 14
2028
99
NM_001323986.1
NP_001310915.1
12 - 13
1935
99
NM_001323987.1
NP_001310916.1
7 - 8
1037
99
NM_001323988.1
NP_001310917.1
10 - 11
1285
99
NM_001323989.1
NP_001310918.1
8 - 9
1133
99
NM_001323990.1
NP_001310919.1
9 - 10
1497
99
NM_001323991.1
NP_001310920.1
8 - 9
1444
99
NM_001323992.1
NP_001310921.1
10 - 11
1277
99
NM_001323993.1
NP_001310922.1
10 - 11
1591
99
NM_001323994.1
NP_001310923.1
7 - 8
1046
99
NM_001323995.1
NP_001310924.1
10 - 11
1261
99
NM_001323996.1
NP_001310925.1
10 - 11
1596
99
NM_001323997.1
NP_001310926.1
10 - 11
1280
99
NM_001323998.1
NP_001310927.1
10 - 11
1572
99
NM_001323999.1
NP_001310928.1
10 - 11
1569
99
NM_001324000.1
NP_001310929.1
8 - 9
1441
99
NM_001324001.1
NP_001310930.1
7 - 8
1348
99
NM_001324002.1
NP_001310931.1
7 - 8
1348
99
NM_001324003.1
NP_001310932.1
9 - 10
1497
99
NM_001324004.1
NP_001310933.1
7 - 8
1037
99
NM_001324005.1
NP_001310934.1
10 - 11
1572
99
NM_001324006.1
NP_001310935.1
11 - 12
1319
99
NM_001324007.1
NP_001310936.1
10 - 11
1555
99
NM_001324008.1
NP_001310937.1
8 - 9
1095
99
NM_001324009.1
NP_001310938.1
8 - 9
1406
99
NM_001324010.1
NP_001310939.1
4 - 5
710
99
NM_001324011.1
NP_001310940.1
9 - 10
1168
99
NM_001324012.1
NP_001310941.1
5 - 6
803
99
NM_001324013.1
NP_001310942.1
8 - 9
1377
99
NM_001903.4
NP_001894.2
13 - 14
2028
99
GenBank mRNA
AK296708.1
-
13 - 14
1836
99
AK300439.1
-
12 - 13
1799
99
AK304267.1
-
13 - 14
1923
99
BC000385.2
-
13 - 14
1966
2821111
99
BC031262.1
-
7 - 8
927
5274351
99
D13866.1
-
13 - 14
1967
99
D14705.1
-
13 - 14
1992
99
HQ589335.1
-
7 - 8
1182
99
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Mammalian Gene Collection
Detect Genomic DNA
Recommended Assay
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene CTNNA1
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
actin filament organization
cell adhesion
establishment or maintenance of cell polarity
negative regulation of neuroblast proliferation
aging
male gonad development
gap junction assembly
axon regeneration
adherens junction organization
cellular protein localization
odontogenesis of dentin-containing tooth
apical junction assembly
response to estrogen
positive regulation of smoothened signaling pathway
positive regulation of muscle cell differentiation
protein heterooligomerization
cellular response to indole-3-methanol
epithelial cell-cell adhesion
negative regulation of cell motility
negative regulation of integrin-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
RNA binding
structural molecule activity
protein binding
beta-catenin binding
vinculin binding
gamma-catenin binding
cadherin binding
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0