Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07288002_g1
          See other CTNNA1 GE Assays ›
          Gene Symbol:
          CTNNA1
          Gene Name:
          catenin alpha 1
          Gene Aliases:
          CAP102, MDPT2
          Chromosome Location:
          Chr.5: 138753386 - 138935034 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07288002_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTNNA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290307.2 NP_001277236.1 15 - 16 2320 100
          NM_001290309.2 NP_001277238.1 14 - 15 2124 100
          NM_001290310.2 NP_001277239.1 15 - 16 2157 100
          NM_001290312.1 NP_001277241.1 9 - 10 1640 100
          NM_001323982.1 NP_001310911.1 16 - 17 2790 100
          NM_001323983.1 NP_001310912.1 15 - 16 2472 100
          NM_001323984.1 NP_001310913.1 16 - 17 2338 100
          NM_001323985.1 NP_001310914.1 15 - 16 2320 100
          NM_001323986.1 NP_001310915.1 14 - 15 2227 100
          NM_001323987.1 NP_001310916.1 9 - 10 1329 100
          NM_001323988.1 NP_001310917.1 12 - 13 1577 100
          NM_001323989.1 NP_001310918.1 10 - 11 1425 100
          NM_001323990.1 NP_001310919.1 11 - 12 1789 100
          NM_001323991.1 NP_001310920.1 10 - 11 1736 100
          NM_001323992.1 NP_001310921.1 12 - 13 1569 100
          NM_001323993.1 NP_001310922.1 12 - 13 1883 100
          NM_001323994.1 NP_001310923.1 9 - 10 1338 100
          NM_001323995.1 NP_001310924.1 12 - 13 1553 100
          NM_001323996.1 NP_001310925.1 12 - 13 1888 100
          NM_001323997.1 NP_001310926.1 12 - 13 1572 100
          NM_001323998.1 NP_001310927.1 12 - 13 1864 100
          NM_001323999.1 NP_001310928.1 12 - 13 1861 100
          NM_001324000.1 NP_001310929.1 10 - 11 1733 100
          NM_001324001.1 NP_001310930.1 9 - 10 1640 100
          NM_001324002.1 NP_001310931.1 9 - 10 1640 100
          NM_001324003.1 NP_001310932.1 11 - 12 1789 100
          NM_001324004.1 NP_001310933.1 9 - 10 1329 100
          NM_001324005.1 NP_001310934.1 12 - 13 1864 100
          NM_001324006.1 NP_001310935.1 13 - 14 1611 100
          NM_001324007.1 NP_001310936.1 12 - 13 1847 100
          NM_001324008.1 NP_001310937.1 10 - 11 1387 100
          NM_001324009.1 NP_001310938.1 10 - 11 1698 100
          NM_001324010.1 NP_001310939.1 6 - 7 1002 100
          NM_001324011.1 NP_001310940.1 11 - 12 1460 100
          NM_001324012.1 NP_001310941.1 7 - 8 1095 100
          NM_001324013.1 NP_001310942.1 10 - 11 1669 100
          NM_001903.4 NP_001894.2 15 - 16 2320 100
          GenBank mRNA AK094151.1 - 12 - 13 1529 100
          AK296708.1 - 15 - 16 2128 100
          AK300439.1 - 14 - 15 2091 100
          AK304267.1 - 15 - 16 2215 100
          BC000385.2 - 15 - 16 2258 2821111 100
          BC031262.1 - 9 - 10 1219 5274351 100
          D13866.1 - 15 - 16 2259 100
          D14705.1 - 15 - 16 2284 100
          HQ589335.1 - 9 - 10 1474 100
          L23805.1 - 14 - 15 2200 100
          U03100.1 - 14 - 15 2200 100

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CTNNA1

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          actin filament organization
          cell adhesion
          establishment or maintenance of cell polarity
          negative regulation of neuroblast proliferation
          aging
          male gonad development
          gap junction assembly
          axon regeneration
          adherens junction organization
          cellular protein localization
          odontogenesis of dentin-containing tooth
          apical junction assembly
          response to estrogen
          positive regulation of smoothened signaling pathway
          positive regulation of muscle cell differentiation
          protein heterooligomerization
          cellular response to indole-3-methanol
          epithelial cell-cell adhesion
          negative regulation of cell motility
          negative regulation of integrin-mediated signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          RNA binding
          structural molecule activity
          protein binding
          beta-catenin binding
          vinculin binding
          gamma-catenin binding
          cadherin binding
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0