Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07288388_m1
          See other CCDC24 GE Assays ›
          Gene Symbol:
          CCDC24
          Gene Name:
          coiled-coil domain containing 24
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 43991247 - 43996528 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07288388_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCDC24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152499.2 NP_689712.1 5 - 6 664 83
          XM_017000423.1 XP_016855912.1 4 - 5 878 83
          XM_017000424.1 XP_016855913.1 4 - 5 1015 83
          XM_017000425.1 XP_016855914.1 4 - 5 878 83
          XM_017000426.1 XP_016855915.1 4 - 5 785 83
          XM_017000427.1 XP_016855916.1 5 - 6 782 83
          XM_017000428.1 XP_016855917.1 5 - 6 779 83
          XM_017000429.1 XP_016855918.1 4 - 5 770 83
          XM_017000430.1 XP_016855919.1 5 - 6 779 83
          XM_017000431.1 XP_016855920.1 5 - 6 671 83
          XM_017000432.1 XP_016855921.1 4 - 5 854 83
          XM_017000433.1 XP_016855922.1 5 - 6 635 83
          XM_017000434.1 XP_016855923.1 5 - 6 635 83
          XM_017000435.1 XP_016855924.1 4 - 5 623 83
          XM_017000436.1 XP_016855925.1 4 - 5 623 83
          XM_017000437.1 XP_016855926.1 5 - 6 581 83
          XM_017000438.1 XP_016855927.1 5 - 6 566 83
          XM_017000439.1 XP_016855928.1 5 - 6 542 83
          XM_017000440.1 XP_016855929.1 5 - 6 527 83
          XM_017000441.1 XP_016855930.1 5 - 6 527 83
          XM_017000442.1 XP_016855931.1 4 - 5 407 83
          XM_017000445.1 XP_016855934.1 3 - 4 279 83
          XM_017000446.1 XP_016855935.1 3 - 4 357 83
          GenBank mRNA AY487420.1 - 4 - 5 385 83
          BC033868.1 - 5 - 6 664 5207706 83
          BC047313.1 - 4 - 5 861 83
          BX641142.1 - 3 - 4 329 83

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCDC24

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0