Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs07289382_g1
See other R3HCC1L GE Assays ›
Gene Symbol:
R3HCC1L
Gene Name:
R3H domain and coiled-coil containing 1 like
Gene Aliases:
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location:
Chr.10: 98134608 - 98244897 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs07289382_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: R3HCC1L
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001256619.1
NP_001243548.1
7 - 8
2347
79
NM_001256620.1
NP_001243549.1
5 - 6
2254
79
NM_001256621.1
NP_001243550.1
4 - 5
451
79
NM_014472.4
NP_055287.4
5 - 6
2250
79
NM_138469.2
NP_612478.2
6 - 7
2347
79
XM_011539639.1
XP_011537941.1
7 - 8
2604
79
XM_011539640.1
XP_011537942.1
8 - 9
2409
79
XM_011539641.1
XP_011537943.1
7 - 8
2361
79
XM_011539642.1
XP_011537944.1
8 - 9
2701
79
XM_011539643.1
XP_011537945.1
6 - 7
2288
79
XM_011539644.1
XP_011537946.1
6 - 7
2292
79
XM_011539645.2
XP_011537947.1
6 - 7
2535
79
XM_011539646.1
XP_011537948.1
5 - 6
2167
79
XM_011539647.1
XP_011537949.1
5 - 6
2169
79
XM_011539648.1
XP_011537950.1
4 - 5
2042
79
XM_011539649.1
XP_011537951.1
6 - 7
2562
79
XM_017016074.1
XP_016871563.1
6 - 7
3500
79
XM_017016075.1
XP_016871564.1
3 - 4
2000
79
XM_017016076.1
XP_016871565.1
6 - 7
2294
79
XM_017016077.1
XP_016871566.1
8 - 9
2457
79
XM_017016078.1
XP_016871567.1
7 - 8
2379
79
XM_017016079.1
XP_016871568.1
7 - 8
2393
79
XM_017016080.1
XP_016871569.1
5 - 6
2299
79
XM_017016081.1
XP_016871570.1
6 - 7
2381
79
XM_017016082.1
XP_016871571.1
4 - 5
2173
79
XM_017016083.1
XP_016871572.1
5 - 6
2189
79
GenBank mRNA
AF525304.1
-
7 - 8
2300
79
AK058187.1
-
1 - 2
41
79
AK094479.1
-
2 - 3
761
79
BC009231.2
-
1 - 2
176
79
BC047908.1
-
4 - 5
397
79
BC098175.1
-
2 - 3
194
79
BC098268.1
-
6 - 7
2288
40026913
79
BC099848.1
-
5 - 6
2191
40026916
79
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene R3HCC1L
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0