Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07289818_m1
          See other PTGER3 GE Assays ›
          Gene Symbol:
          PTGER3
          Gene Name:
          prostaglandin E receptor 3
          Gene Aliases:
          EP3, EP3-I, EP3-II, EP3-III, EP3-IV, EP3-VI, EP3e, PGE2-R
          Chromosome Location:
          Chr.1: 70852353 - 71047814 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07289818_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTGER3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198719.1 NP_942012.1 3 - 4 1400 77
          GenBank mRNA AK315825.1 - 3 - 4 1328 77
          D38297.1 - 3 - 4 1400 77
          L26976.1 - 3 - 4 1308 77
          L27490.1 - 3 - 4 1364 77
          S68874.1 - 1 - 2 92 77
          U13218.1 - 3 - 4 1225 77
          X83858.1 - 3 - 4 1376 77

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PTGER3

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          G protein-coupled receptor signaling pathway
          adenylate cyclase-activating G protein-coupled receptor signaling pathway
          phospholipase C-activating G protein-coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          female pregnancy
          cell death
          response to xenobiotic stimulus
          positive regulation of gene expression
          negative regulation of norepinephrine secretion
          intestine smooth muscle contraction
          urinary bladder smooth muscle contraction
          positive regulation of fever generation
          response to estradiol
          response to progesterone
          positive regulation of vasoconstriction
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of insulin secretion
          maternal process involved in parturition
          negative regulation of gastric acid secretion
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          negative regulation of platelet aggregation
          negative regulation of potassium ion transmembrane transport
          response to salt
          negative regulation of calcium ion transmembrane transport
          negative regulation of gastrin-induced gastric acid secretion
          positive regulation of smooth muscle contraction involved in micturition
          cellular response to forskolin
          positive regulation of inhibitory G protein-coupled receptor phosphorylation
          negative regulation of circadian sleep/wake cycle, wakefulness
          positive regulation of myofibroblast contraction
          positive regulation of colon smooth muscle contraction
          positive regulation of gastric mucosal blood circulation
          prostaglandin receptor internalization
          positive regulation of neutrophil extravasation
          negative regulation of forebrain neuron differentiation
          prostaglandin E receptor activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0