Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07289828_g1
          See other GGTLC2 GE Assays ›
          Gene Symbol:
          GGTLC2 GGT2 GGTLC1 LOC102724197
          Gene Name:
          gamma-glutamyltransferase light chain 2
          gamma-glutamyltransferase 2
          gamma-glutamyltransferase light chain 1
          inactive gamma-glutamyltranspeptidase 2-like
          Gene Aliases:
          GGTL4
          GGT, GGT 2
          GGTL6, GGTLA3, GGTLA4, dJ831C21.1, dJ831C21.2
          GGT2
          Chromosome Location:
          Chr.22: 22644620 - 22647898 on Build GRCh38
          Chromosome Location:
          Chr.22: 21207972 - 21282982 on Build GRCh38
          Chromosome Location:
          Chr.20: 23985053 - 23989134 on Build GRCh38
          Chromosome Location:
          Chr.22|NT_187386: 65299 - 86956 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          Assay ID Hs07289828_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GGTLC2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282879.1 NP_001269808.1 3 - 4 367 219
          NM_199127.2 NP_954578.2 2 - 3 339 117
          XM_005261815.3 XP_005261872.2 3 - 4 396 219
          XM_005261817.2 XP_005261874.1 3 - 4 371 219
          XM_005261818.2 XP_005261875.1 3 - 4 396 117
          XM_011530497.1 XP_011528799.1 3 - 4 479 219
          GenBank mRNA BC069534.1 - 2 - 3 305 7262125 117
          EL736203.1 - 2 - 3 305 117
          X98922.1 - 2 - 3 340 219
          Gene Symbol: GGT2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005261890.4 XP_005261947.1 6 - 7 890 219
          XM_006724392.3 XP_006724455.1 10 - 11 1598 219
          XM_006724395.3 XP_006724458.1 7 - 8 1050 219
          XM_006724396.2 XP_006724459.1 3 - 4 428 219
          XM_017029179.1 XP_016884668.1 11 - 12 2279 219
          XM_017029180.1 XP_016884669.1 10 - 11 1737 219
          XM_017029181.1 XP_016884670.1 10 - 11 2175 219
          XM_017029183.1 XP_016884672.1 6 - 7 866 219
          XM_017029184.1 XP_016884673.1 3 - 4 438 219
          Gene Symbol: GGTLC1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005260863.4 XP_005260920.1 3 - 4 865 219
          XM_005260864.4 XP_005260921.1 4 - 5 1018 219
          XM_011529391.2 XP_011527693.1 3 - 4 432 219
          Gene Symbol: LOC102724197
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006724921.3 XP_006724984.1 10 - 11 2041 219
          XM_006724924.3 XP_006724987.1 9 - 10 1883 219
          XM_006724928.3 XP_006724991.1 5 - 6 1079 219
          XM_006724929.2 XP_006724992.1 3 - 4 428 219
          XM_017030133.1 XP_016885622.1 11 - 12 1606 219
          XM_017030134.1 XP_016885623.1 11 - 12 1609 219
          XM_017030135.1 XP_016885624.1 11 - 12 2034 219
          XM_017030136.1 XP_016885625.1 12 - 13 2076 219
          XM_017030137.1 XP_016885626.1 12 - 13 2073 219
          XM_017030138.1 XP_016885627.1 12 - 13 1596 219
          XM_017030139.1 XP_016885628.1 12 - 13 1599 219
          XM_017030140.1 XP_016885629.1 13 - 14 2066 219
          XM_017030141.1 XP_016885630.1 13 - 14 2063 219
          XM_017030142.1 XP_016885631.1 11 - 12 1907 219
          XM_017030144.1 XP_016885633.1 6 - 7 970 219
          XM_017030145.1 XP_016885634.1 7 - 8 1603 219

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GGTLC2
          Click here to find siRNAs for Gene GGT2
          Click here to find siRNAs for Gene GGTLC1
          Click here to find siRNAs for Gene LOC102724197

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          leukotriene biosynthetic process
          glutathione biosynthetic process
          xenobiotic metabolic process
          zymogen activation
          glutathione hydrolase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0