Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07290489_m1
          See other ALG13 GE Assays ›
          Gene Symbol:
          ALG13
          Gene Name:
          ALG13, UDP-N-acetylglucosaminyltransferase subunit
          Gene Aliases:
          CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
          Chromosome Location:
          Chr.X: 111680741 - 111764164 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07290489_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ALG13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099922.2 NP_001093392.1 2 - 3 346 78
          NM_001168385.2 NP_001161857.1 2 - 3 346 78
          NM_001257230.1 NP_001244159.1 2 - 3 242 78
          NM_001257234.1 NP_001244163.1 2 - 3 315 78
          NM_001257235.2 NP_001244164.1 2 - 3 315 78
          NM_001257237.1 NP_001244166.1 2 - 3 419 78
          NM_001257239.2 NP_001244168.1 2 - 3 334 78
          NM_001257240.2 NP_001244169.1 2 - 3 419 78
          NM_001324291.1 NP_001311220.1 2 - 3 242 78
          NM_001324292.1 NP_001311221.1 2 - 3 346 78
          NM_001324293.1 NP_001311222.1 2 - 3 419 78
          NM_001324294.1 NP_001311223.1 2 - 3 230 78
          NM_018466.5 NP_060936.1 2 - 3 346 78
          NR_033125.2 2 - 3 242 78
          XM_006724693.2 XP_006724756.1 2 - 3 719 78
          XM_006724697.3 XP_006724760.2 2 - 3 619 78
          XM_006724698.3 XP_006724761.2 2 - 3 744 78
          XM_011531028.2 XP_011529330.1 2 - 3 720 78
          XM_011531030.1 XP_011529332.1 2 - 3 718 78
          XM_011531031.1 XP_011529333.1 2 - 3 419 78
          XM_011531032.1 XP_011529334.1 2 - 3 225 78
          XM_011531033.2 XP_011529335.1 2 - 3 265 78
          XM_017029846.1 XP_016885335.1 2 - 3 721 78
          XM_017029847.1 XP_016885336.1 2 - 3 720 78
          XM_017029848.1 XP_016885337.1 2 - 3 718 78
          XM_017029849.1 XP_016885338.1 2 - 3 719 78
          XM_017029850.1 XP_016885339.1 2 - 3 718 78
          XM_017029851.1 XP_016885340.1 2 - 3 721 78
          GenBank mRNA AF220051.1 - 2 - 3 277 78
          AK300394.1 - 2 - 3 184 78
          AK302729.1 - 2 - 3 287 78
          AK304712.1 - 2 - 3 236 78
          AK312229.1 - 2 - 3 285 78
          AK316306.1 - 2 - 3 288 78
          AK316522.1 - 2 - 3 363 78
          AL524204.3 - 2 - 3 165 78
          BC005336.1 - 2 - 3 279 3996922 78
          BE747995.1 - 2 - 3 275 78
          BG621042.1 - 1 - 2 129 78
          CB268871.1 - 2 - 3 293 78
          DA830058.1 - 2 - 3 410 78
          DB292609.1 - 2 - 3 419 78
          DC400909.1 - 2 - 3 230 78
          DN992019.1 - 2 - 3 414 78

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK300394.1, AK304712.1, AK316306.1, AK316522.1, NM_001257230.1, NM_001257234.1, NM_001257235.2, NM_001257237.1, NM_001257239.2, NM_001257240.2, NM_001324291.1, NM_001324293.1, NM_001324294.1, XM_006724697.3, XM_006724698.3, XM_011531031.1, XM_011531032.1, XM_011531033.2, XM_017029851.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ALG13

          Gene Ontology Categories:

          Function(s) Process(es)

          dolichol-linked oligosaccharide biosynthetic process
          proteolysis
          RNA binding
          N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
          thiol-dependent deubiquitinase

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0