Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07290569_m1
          See other CARD8 GE Assays ›
          Gene Symbol:
          CARD8
          Gene Name:
          caspase recruitment domain family member 8
          Gene Aliases:
          CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
          Chromosome Location:
          Chr.19: 48203148 - 48256269 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07290569_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CARD8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001184900.1 NP_001171829.1 1 - 2 102 68
          NM_001184902.1 NP_001171831.1 4 - 5 465 68
          NM_001184903.1 NP_001171832.1 2 - 3 199 68
          NM_001184904.1 NP_001171833.1 3 - 4 354 68
          NR_033679.1 1 - 2 102 68
          XM_006723091.3 XP_006723154.1 3 - 4 313 68
          XM_006723092.3 XP_006723155.1 5 - 6 851 68
          XM_006723093.3 XP_006723156.1 3 - 4 634 68
          XM_006723094.3 XP_006723157.1 3 - 4 296 68
          XM_006723098.3 XP_006723161.1 3 - 4 313 68
          XM_006723099.3 XP_006723162.1 2 - 3 199 68
          XM_006723106.3 XP_006723169.1 3 - 4 645 68
          XM_006723107.3 XP_006723170.1 3 - 4 296 68
          XM_006723110.3 XP_006723173.1 4 - 5 783 68
          XM_011526641.2 XP_011524943.1 6 - 7 1158 68
          XM_011526642.2 XP_011524944.1 5 - 6 1053 68
          XM_011526643.2 XP_011524945.1 4 - 5 740 68
          XM_011526644.2 XP_011524946.1 2 - 3 325 68
          XM_011526646.2 XP_011524948.1 4 - 5 749 68
          XM_011526647.2 XP_011524949.1 3 - 4 638 68
          XM_011526648.2 XP_011524950.1 4 - 5 758 68
          XM_011526650.2 XP_011524952.1 3 - 4 296 68
          XM_011526653.2 XP_011524955.1 4 - 5 788 68
          XM_017026482.1 XP_016881971.1 3 - 4 641 68
          XM_017026485.1 XP_016881974.1 3 - 4 296 68
          XM_017026486.1 XP_016881975.1 2 - 3 199 68
          XM_017026490.1 XP_016881979.1 3 - 4 296 68
          XM_017026491.1 XP_016881980.1 3 - 4 296 68
          XM_017026495.1 XP_016881984.1 3 - 4 296 68
          GenBank mRNA AI040349.1 - 1 - 2 281 68
          AK297045.1 - 2 - 3 185 68
          BC034564.1 - 3 - 4 301 68
          BX648568.1 - 3 - 4 297 68
          EU118120.1 - 1 - 2 102 68
          EU118121.1 - 1 - 2 102 68

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the EU118121.1, XM_006723098.3, XM_006723099.3, XM_011526646.2, XM_011526647.2, XM_011526648.2, XM_011526653.2, XM_017026486.1, XM_017026495.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          protein binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          NACHT domain binding
          protein homodimerization activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0