Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00431636_m1
          See other CHRNA7 GE Assays ›
          Gene Symbol:
          Chrna7
          Gene Name:
          cholinergic receptor, nicotinic, alpha polypeptide 7
          Gene Aliases:
          Acra7, alpha7, nAchR
          Chromosome Location:
          Chr.7: 63098692 - 63212526 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00431636_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrna7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007390.3 NP_031416.3 5 - 6 479 77
          GenBank mRNA AF225980.1 - 5 - 6 469 77
          AF325348.1 - 4 - 5 276 77
          AK157809.1 - 5 - 6 476 77
          AY574266.1 - 5 - 6 429 77
          L37663.1 - 5 - 6 479 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003403.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Mouse Alzheimer's Disease Inventoried Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chrna7

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of heart rate involved in baroreceptor response to decreased systemic arterial blood pressure
          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          endocytosis
          signal transduction
          synaptic transmission, cholinergic
          memory
          positive regulation of cell population proliferation
          associative learning
          regulation of norepinephrine secretion
          neuronal action potential
          flagellated sperm motility
          response to food
          regulation of synaptic transmission, dopaminergic
          negative regulation of interleukin-1 beta production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          ion transmembrane transport
          response to nicotine
          behavioral response to nicotine
          T cell activation
          B cell activation
          regulation of membrane potential
          ovulation cycle
          response to ethanol
          positive regulation of angiogenesis
          behavioral response to ethanol
          regulation of inflammatory response
          negative regulation of inflammatory response
          cognition
          generation of ovulation cycle rhythm
          cation transmembrane transport
          amyloid-beta binding
          ion channel activity
          extracellular ligand-gated ion channel activity
          protein binding
          adenylate cyclase binding
          acetylcholine receptor activity
          toxic substance binding
          chloride channel regulator activity
          protein kinase binding
          acetylcholine-gated cation-selective channel activity
          acetylcholine binding
          protein homodimerization activity
          ATPase binding
          scaffold protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700029051.mm.1 TC0700003403.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029051.mm.1 NM_007390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029051.mm.1 NM_007390.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0